Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 147878693-147878966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf46
Entrez Gene ID
389336
Gene Name
chromosome 5 open reading frame 46
Gene Aliases
SSSP1
UniGene
Hs.660038
Gene Chromosome Location
Chr.5: 147850463-147906594 on Build GRCh38
Transcripts
XM_017009459.1 NM_054023.4

Gene Symbol
SCGB3A2
Entrez Gene ID
117156
Gene Name
secretoglobin family 3A member 2
Gene Aliases
LU103, PNSP1, UGRP1, pnSP-1
UniGene
Hs.483765
Gene Chromosome Location
Chr.5: 147878711-147882193 on Build GRCh38
Transcripts
XM_017009459.1 NM_054023.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5026432 c.27G>A p.L9L Chr.5: 147878830-147878830 on Build GRCh38
1224785 c.32C>T p.T11I Chr.5: 147878835-147878835 on Build GRCh38
3612136 c.34A>G p.I12V Chr.5: 147878837-147878837 on Build GRCh38
4585599 c.39C>T p.S13S Chr.5: 147878842-147878842 on Build GRCh38
5351167 c.25C>A p.L9M Chr.5: 147878828-147878828 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747226682 Chr.5: 147878878-147878878 on Build GRCh38
rs749205893 Chr.5: 147878889-147878889 on Build GRCh38
rs777530393 Chr.5: 147878854-147878854 on Build GRCh38
rs758419757 Chr.5: 147878862-147878862 on Build GRCh38
rs577934971 Chr.5: 147878742-147878743 on Build GRCh38
rs1039560769 Chr.5: 147878819-147878819 on Build GRCh38
rs780272130 Chr.5: 147878875-147878875 on Build GRCh38
rs561609651 Chr.5: 147878718-147878718 on Build GRCh38
rs765849502 Chr.5: 147878775-147878775 on Build GRCh38
rs773037092 Chr.5: 147878887-147878887 on Build GRCh38
rs2278376 Chr.5: 147878932-147878932 on Build GRCh38
rs147682862 Chr.5: 147878821-147878821 on Build GRCh38
rs372563565 Chr.5: 147878923-147878923 on Build GRCh38
rs114767590 Chr.5: 147878755-147878755 on Build GRCh38
rs546898562 Chr.5: 147878804-147878804 on Build GRCh38
rs372667519 Chr.5: 147878778-147878780 on Build GRCh38
rs779124435 Chr.5: 147878840-147878840 on Build GRCh38
rs370711811 Chr.5: 147878803-147878803 on Build GRCh38
rs757889024 Chr.5: 147878837-147878837 on Build GRCh38
rs764587619 Chr.5: 147878811-147878811 on Build GRCh38
rs149064485 Chr.5: 147878728-147878729 on Build GRCh38
rs757938827 Chr.5: 147878821-147878822 on Build GRCh38
rs759103396 Chr.5: 147878791-147878791 on Build GRCh38
rs200326093 Chr.5: 147878908-147878908 on Build GRCh38
rs564837835 Chr.5: 147878939-147878939 on Build GRCh38
rs368131235 Chr.5: 147878799-147878799 on Build GRCh38
rs1012931828 Chr.5: 147878912-147878912 on Build GRCh38
rs746029169 Chr.5: 147878841-147878841 on Build GRCh38
rs1049504395 Chr.5: 147878781-147878781 on Build GRCh38
rs181768391 Chr.5: 147878754-147878754 on Build GRCh38
rs942403789 Chr.5: 147878817-147878817 on Build GRCh38
rs773092660 Chr.5: 147878762-147878762 on Build GRCh38
rs386693354 Chr.5: 147878932-147878933 on Build GRCh38
rs75055284 Chr.5: 147878933-147878933 on Build GRCh38
rs751123914 Chr.5: 147878788-147878788 on Build GRCh38
rs543232043 Chr.5: 147878736-147878736 on Build GRCh38
rs1002939453 Chr.5: 147878894-147878894 on Build GRCh38
rs770919743 Chr.5: 147878891-147878891 on Build GRCh38
rs758610324 Chr.5: 147878890-147878890 on Build GRCh38
rs767127545 Chr.5: 147878793-147878793 on Build GRCh38
rs893055956 Chr.5: 147878824-147878824 on Build GRCh38
rs141389076 Chr.5: 147878776-147878778 on Build GRCh38
rs1041562787 Chr.5: 147878854-147878854 on Build GRCh38
rs986542969 Chr.5: 147878778-147878778 on Build GRCh38
rs374637255 Chr.5: 147878763-147878763 on Build GRCh38
rs918308806 Chr.5: 147878739-147878739 on Build GRCh38
rs769599414 Chr.5: 147878883-147878883 on Build GRCh38
rs762916628 Chr.5: 147878765-147878765 on Build GRCh38
rs144571393 Chr.5: 147878900-147878900 on Build GRCh38

View Full Product Details