Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 112780907-112781154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMOT
Entrez Gene ID
154796
Gene Name
angiomotin
Gene Aliases
-
UniGene
Hs.528051
Gene Chromosome Location
Chr.X: 112774877-112840946 on Build GRCh38
Transcripts
NM_001113490.1 NM_133265.2 XM_005262087.1 XM_005262090.1 XM_011530875.2 XM_017029289.1

SNP Detail

SNP ID Chromosome Location
rs763760828 Chr.X: 112781021-112781021 on Build GRCh38
rs370882819 Chr.X: 112780980-112780980 on Build GRCh38
rs748093114 Chr.X: 112780992-112780992 on Build GRCh38
rs766135042 Chr.X: 112780988-112780988 on Build GRCh38
rs749411318 Chr.X: 112781058-112781058 on Build GRCh38
rs770331835 Chr.X: 112780947-112780947 on Build GRCh38
rs769005431 Chr.X: 112780970-112780970 on Build GRCh38
rs145715476 Chr.X: 112780981-112780981 on Build GRCh38
rs756906269 Chr.X: 112781040-112781040 on Build GRCh38
rs771035290 Chr.X: 112781005-112781005 on Build GRCh38
rs775771819 Chr.X: 112780959-112780959 on Build GRCh38
rs890348269 Chr.X: 112781110-112781110 on Build GRCh38
rs768234510 Chr.X: 112781041-112781041 on Build GRCh38
rs149909515 Chr.X: 112781019-112781019 on Build GRCh38
rs1044761025 Chr.X: 112780984-112780984 on Build GRCh38
rs374222593 Chr.X: 112781036-112781036 on Build GRCh38
rs749322989 Chr.X: 112780962-112780962 on Build GRCh38
rs780642944 Chr.X: 112781050-112781050 on Build GRCh38
rs756592778 Chr.X: 112781045-112781045 on Build GRCh38
rs747409433 Chr.X: 112780930-112780930 on Build GRCh38
rs768805368 Chr.X: 112781076-112781076 on Build GRCh38
rs746405064 Chr.X: 112780945-112780945 on Build GRCh38
rs758101016 Chr.X: 112780928-112780928 on Build GRCh38
rs757833562 Chr.X: 112781020-112781020 on Build GRCh38
rs781600309 Chr.X: 112780936-112780936 on Build GRCh38
rs776575428 Chr.X: 112781037-112781037 on Build GRCh38
rs769380289 Chr.X: 112781011-112781011 on Build GRCh38
rs377505847 Chr.X: 112780967-112780967 on Build GRCh38
rs774548706 Chr.X: 112781126-112781126 on Build GRCh38
rs757518646 Chr.X: 112780932-112780932 on Build GRCh38
rs865963760 Chr.X: 112781006-112781006 on Build GRCh38
rs2286063 Chr.X: 112780929-112780929 on Build GRCh38
rs759455437 Chr.X: 112781004-112781004 on Build GRCh38

View Full Product Details