Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74528045-74528308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUP1
Entrez Gene ID
550
Gene Name
ancient ubiquitous protein 1
Gene Aliases
-
UniGene
Hs.411480
Gene Chromosome Location
Chr.2: 74526648-74529897 on Build GRCh38
Transcripts
NM_181575.4 NR_126510.1 NR_126511.1 XM_005264392.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086100 c.650C>G p.P217R Chr.2: 74528269-74528269 on Build GRCh38
3991463 c.645C>A p.F215L Chr.2: 74528274-74528274 on Build GRCh38
6467422 c.620T>C p.V207A Chr.2: 74528299-74528299 on Build GRCh38
7357133 c.653T>A p.F218Y Chr.2: 74528266-74528266 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs978826583 Chr.2: 74528074-74528074 on Build GRCh38
rs528209132 Chr.2: 74528221-74528221 on Build GRCh38
rs941604748 Chr.2: 74528176-74528176 on Build GRCh38
rs924604418 Chr.2: 74528087-74528087 on Build GRCh38
rs779491856 Chr.2: 74528283-74528283 on Build GRCh38
rs750631652 Chr.2: 74528239-74528239 on Build GRCh38
rs781316727 Chr.2: 74528237-74528237 on Build GRCh38
rs377142567 Chr.2: 74528273-74528273 on Build GRCh38
rs760458647 Chr.2: 74528207-74528207 on Build GRCh38
rs932792743 Chr.2: 74528165-74528165 on Build GRCh38
rs756378752 Chr.2: 74528254-74528254 on Build GRCh38
rs564024289 Chr.2: 74528127-74528127 on Build GRCh38
rs766252276 Chr.2: 74528210-74528210 on Build GRCh38
rs934773091 Chr.2: 74528089-74528089 on Build GRCh38
rs1012713980 Chr.2: 74528242-74528242 on Build GRCh38
rs139159726 Chr.2: 74528205-74528205 on Build GRCh38
rs756146670 Chr.2: 74528084-74528084 on Build GRCh38
rs146230226 Chr.2: 74528098-74528098 on Build GRCh38
rs758972134 Chr.2: 74528140-74528140 on Build GRCh38
rs757362595 Chr.2: 74528236-74528236 on Build GRCh38
rs768998963 Chr.2: 74528278-74528278 on Build GRCh38
rs751535897 Chr.2: 74528231-74528231 on Build GRCh38
rs530385166 Chr.2: 74528131-74528131 on Build GRCh38
rs749687703 Chr.2: 74528262-74528262 on Build GRCh38
rs780368452 Chr.2: 74528260-74528260 on Build GRCh38
rs564019876 Chr.2: 74528215-74528215 on Build GRCh38
rs888665528 Chr.2: 74528169-74528169 on Build GRCh38
rs771553804 Chr.2: 74528198-74528198 on Build GRCh38
rs765241193 Chr.2: 74528224-74528224 on Build GRCh38
rs748665825 Chr.2: 74528286-74528286 on Build GRCh38
rs1053388719 Chr.2: 74528064-74528064 on Build GRCh38
rs1050438053 Chr.2: 74528167-74528167 on Build GRCh38
rs976784963 Chr.2: 74528116-74528116 on Build GRCh38
rs369751295 Chr.2: 74528202-74528202 on Build GRCh38

View Full Product Details