Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30652773-30653046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf136
Entrez Gene ID
221545
Gene Name
chromosome 6 open reading frame 136
Gene Aliases
-
UniGene
Hs.591787
Gene Chromosome Location
Chr.6: 30647039-30653210 on Build GRCh38
Transcripts
NM_001109938.2 NM_001161376.1 NM_145029.3 XM_011514384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574765 c.441G>A p.K147K Chr.6: 30652837-30652837 on Build GRCh38
3251583 c.431C>A p.T144K Chr.6: 30652827-30652827 on Build GRCh38
7326871 c.1394_1395insA p.P466fs*24 Chr.6: 30652818-30652819 on Build GRCh38
7326870 c.422_423insA p.P142fs*24 Chr.6: 30652818-30652819 on Build GRCh38
7236596 c.431C>T p.T144M Chr.6: 30652827-30652827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774413817 Chr.6: 30652886-30652886 on Build GRCh38
rs1050439076 Chr.6: 30652968-30652968 on Build GRCh38
rs577308677 Chr.6: 30652956-30652956 on Build GRCh38
rs758471981 Chr.6: 30652840-30652840 on Build GRCh38
rs546238562 Chr.6: 30653009-30653009 on Build GRCh38
rs554341893 Chr.6: 30652932-30652932 on Build GRCh38
rs754426497 Chr.6: 30653007-30653007 on Build GRCh38
rs774670424 Chr.6: 30652957-30652958 on Build GRCh38
rs747144916 Chr.6: 30652850-30652850 on Build GRCh38
rs761481141 Chr.6: 30652896-30652896 on Build GRCh38
rs767067882 Chr.6: 30652905-30652905 on Build GRCh38
rs749151397 Chr.6: 30652944-30652948 on Build GRCh38
rs142538587 Chr.6: 30652906-30652906 on Build GRCh38
rs749076846 Chr.6: 30652870-30652870 on Build GRCh38
rs775105867 Chr.6: 30652809-30652809 on Build GRCh38
rs552798979 Chr.6: 30652944-30652945 on Build GRCh38
rs966681907 Chr.6: 30652939-30652939 on Build GRCh38
rs775104442 Chr.6: 30652858-30652858 on Build GRCh38
rs935497907 Chr.6: 30652837-30652837 on Build GRCh38
rs755521488 Chr.6: 30652806-30652806 on Build GRCh38
rs368160789 Chr.6: 30652882-30652882 on Build GRCh38
rs757458062 Chr.6: 30652964-30652964 on Build GRCh38
rs562564429 Chr.6: 30653022-30653022 on Build GRCh38
rs961162597 Chr.6: 30653011-30653011 on Build GRCh38
rs201543406 Chr.6: 30652828-30652828 on Build GRCh38
rs369570977 Chr.6: 30652925-30652925 on Build GRCh38
rs375230807 Chr.6: 30652955-30652955 on Build GRCh38
rs769656246 Chr.6: 30652852-30652852 on Build GRCh38
rs993963019 Chr.6: 30653014-30653014 on Build GRCh38
rs753361205 Chr.6: 30652934-30652934 on Build GRCh38
rs1034855741 Chr.6: 30653016-30653016 on Build GRCh38
rs150959328 Chr.6: 30652917-30652917 on Build GRCh38
rs1016820919 Chr.6: 30653010-30653010 on Build GRCh38
rs758490156 Chr.6: 30652951-30652951 on Build GRCh38
rs765800497 Chr.6: 30652938-30652938 on Build GRCh38
rs778064836 Chr.6: 30652846-30652846 on Build GRCh38
rs887798751 Chr.6: 30653006-30653006 on Build GRCh38
rs779372396 Chr.6: 30652819-30652819 on Build GRCh38
rs760195205 Chr.6: 30652912-30652912 on Build GRCh38
rs753140414 Chr.6: 30652950-30652950 on Build GRCh38

View Full Product Details