Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80081725-80081998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC40
Entrez Gene ID
55036
Gene Name
coiled-coil domain containing 40
Gene Aliases
CILD15, FAP172
UniGene
Hs.202542
Gene Chromosome Location
Chr.17: 80036632-80100613 on Build GRCh38
Transcripts
NM_001243342.1 NM_017950.3 XM_011524963.2 XM_011524964.2 XM_011524965.2 XM_017024807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5972475 c.1883C>T p.T628M Chr.17: 80081952-80081952 on Build GRCh38
1290775 c.1743C>T p.S581S Chr.17: 80081726-80081726 on Build GRCh38
3755976 c.1890T>G p.A630A Chr.17: 80081959-80081959 on Build GRCh38
4595799 c.1889C>T p.A630V Chr.17: 80081958-80081958 on Build GRCh38
4623685 c.1927A>G p.N643D Chr.17: 80081996-80081996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765427963 Chr.17: 80081915-80081915 on Build GRCh38
rs772033768 Chr.17: 80081968-80081968 on Build GRCh38
rs200551690 Chr.17: 80081966-80081966 on Build GRCh38
rs765193858 Chr.17: 80081893-80081895 on Build GRCh38
rs376363442 Chr.17: 80081952-80081952 on Build GRCh38
rs765420679 Chr.17: 80081765-80081765 on Build GRCh38
rs35578653 Chr.17: 80081959-80081959 on Build GRCh38
rs530585863 Chr.17: 80081794-80081794 on Build GRCh38
rs1014600453 Chr.17: 80081790-80081790 on Build GRCh38
rs759468416 Chr.17: 80081967-80081967 on Build GRCh38
rs377745657 Chr.17: 80081868-80081868 on Build GRCh38
rs751321015 Chr.17: 80081802-80081802 on Build GRCh38
rs767290092 Chr.17: 80081883-80081883 on Build GRCh38
rs753336334 Chr.17: 80081913-80081913 on Build GRCh38
rs752363930 Chr.17: 80081805-80081805 on Build GRCh38
rs965506812 Chr.17: 80081783-80081783 on Build GRCh38
rs748367580 Chr.17: 80081844-80081844 on Build GRCh38
rs386799737 Chr.17: 80081958-80081959 on Build GRCh38
rs534787961 Chr.17: 80081849-80081849 on Build GRCh38
rs371383125 Chr.17: 80081893-80081893 on Build GRCh38
rs182104049 Chr.17: 80081797-80081797 on Build GRCh38
rs948532763 Chr.17: 80081876-80081876 on Build GRCh38
rs754801803 Chr.17: 80081803-80081803 on Build GRCh38
rs887097108 Chr.17: 80081946-80081946 on Build GRCh38
rs763700391 Chr.17: 80081912-80081912 on Build GRCh38
rs375215801 Chr.17: 80081864-80081864 on Build GRCh38
rs61749058 Chr.17: 80081958-80081958 on Build GRCh38
rs767093580 Chr.17: 80081804-80081804 on Build GRCh38
rs761918200 Chr.17: 80081747-80081747 on Build GRCh38
rs886053534 Chr.17: 80081965-80081965 on Build GRCh38
rs770932464 Chr.17: 80081963-80081963 on Build GRCh38
rs769950790 Chr.17: 80081848-80081848 on Build GRCh38
rs756747733 Chr.17: 80081920-80081920 on Build GRCh38
rs752551759 Chr.17: 80081892-80081892 on Build GRCh38
rs755600220 Chr.17: 80081943-80081943 on Build GRCh38
rs748572237 Chr.17: 80081953-80081953 on Build GRCh38
rs199791020 Chr.17: 80081837-80081837 on Build GRCh38
rs773232601 Chr.17: 80081796-80081796 on Build GRCh38
rs571288423 Chr.17: 80081929-80081929 on Build GRCh38
rs373309118 Chr.17: 80081829-80081829 on Build GRCh38
rs777329186 Chr.17: 80081817-80081817 on Build GRCh38
rs759273764 Chr.17: 80081872-80081872 on Build GRCh38
rs752125203 Chr.17: 80081930-80081930 on Build GRCh38
rs747335744 Chr.17: 80081828-80081828 on Build GRCh38
rs748784362 Chr.17: 80081824-80081824 on Build GRCh38
rs755826267 Chr.17: 80081808-80081808 on Build GRCh38
rs577764470 Chr.17: 80081825-80081825 on Build GRCh38
rs1006029303 Chr.17: 80081754-80081755 on Build GRCh38
rs113887654 Chr.17: 80081836-80081836 on Build GRCh38
rs899000567 Chr.17: 80081910-80081910 on Build GRCh38
rs766278175 Chr.17: 80081798-80081798 on Build GRCh38
rs369273325 Chr.17: 80081866-80081866 on Build GRCh38

View Full Product Details