Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 88040077-88040344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf29
Entrez Gene ID
91298
Gene Name
chromosome 12 open reading frame 29
Gene Aliases
-
UniGene
Hs.591009
Gene Chromosome Location
Chr.12: 88035491-88050160 on Build GRCh38
Transcripts
NM_001009894.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6414877 c.140C>T p.A47V Chr.12: 88040197-88040197 on Build GRCh38
4431310 c.175G>A p.D59N Chr.12: 88040232-88040232 on Build GRCh38
6614161 c.91-10delT p.? Chr.12: 88040138-88040138 on Build GRCh38
5803511 c.116C>G p.T39S Chr.12: 88040173-88040173 on Build GRCh38
1739948 c.152G>A p.S51N Chr.12: 88040209-88040209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759133448 Chr.12: 88040290-88040290 on Build GRCh38
rs780376396 Chr.12: 88040232-88040232 on Build GRCh38
rs367808948 Chr.12: 88040115-88040115 on Build GRCh38
rs370635197 Chr.12: 88040113-88040113 on Build GRCh38
rs1021998026 Chr.12: 88040198-88040198 on Build GRCh38
rs766696689 Chr.12: 88040174-88040175 on Build GRCh38
rs754396582 Chr.12: 88040184-88040184 on Build GRCh38
rs766905258 Chr.12: 88040306-88040306 on Build GRCh38
rs372673173 Chr.12: 88040209-88040209 on Build GRCh38
rs769697563 Chr.12: 88040257-88040257 on Build GRCh38
rs758928030 Chr.12: 88040145-88040145 on Build GRCh38
rs761946280 Chr.12: 88040129-88040129 on Build GRCh38
rs773324273 Chr.12: 88040123-88040123 on Build GRCh38
rs148273000 Chr.12: 88040173-88040173 on Build GRCh38
rs751129541 Chr.12: 88040138-88040138 on Build GRCh38
rs766046913 Chr.12: 88040133-88040133 on Build GRCh38
rs778948869 Chr.12: 88040109-88040109 on Build GRCh38
rs773127867 Chr.12: 88040260-88040260 on Build GRCh38
rs923840752 Chr.12: 88040294-88040294 on Build GRCh38
rs1008362999 Chr.12: 88040144-88040144 on Build GRCh38
rs757772265 Chr.12: 88040185-88040185 on Build GRCh38
rs545981199 Chr.12: 88040262-88040262 on Build GRCh38
rs756541240 Chr.12: 88040174-88040174 on Build GRCh38
rs767219478 Chr.12: 88040154-88040154 on Build GRCh38
rs747171535 Chr.12: 88040249-88040249 on Build GRCh38
rs762980594 Chr.12: 88040124-88040124 on Build GRCh38
rs769458510 Chr.12: 88040122-88040122 on Build GRCh38
rs538745901 Chr.12: 88040263-88040263 on Build GRCh38
rs377345842 Chr.12: 88040279-88040279 on Build GRCh38
rs761539515 Chr.12: 88040114-88040125 on Build GRCh38
rs776594494 Chr.12: 88040107-88040110 on Build GRCh38
rs752268951 Chr.12: 88040160-88040160 on Build GRCh38
rs770998752 Chr.12: 88040276-88040276 on Build GRCh38
rs772854622 Chr.12: 88040128-88040131 on Build GRCh38
rs745982661 Chr.12: 88040205-88040205 on Build GRCh38
rs769682828 Chr.12: 88040128-88040129 on Build GRCh38
rs764359263 Chr.12: 88040179-88040179 on Build GRCh38
rs763335811 Chr.12: 88040138-88040139 on Build GRCh38
rs141391830 Chr.12: 88040204-88040204 on Build GRCh38
rs761694704 Chr.12: 88040118-88040118 on Build GRCh38
rs774381102 Chr.12: 88040277-88040277 on Build GRCh38
rs763266905 Chr.12: 88040241-88040241 on Build GRCh38
rs751702625 Chr.12: 88040241-88040243 on Build GRCh38
rs766743394 Chr.12: 88040305-88040305 on Build GRCh38
rs1018852969 Chr.12: 88040150-88040150 on Build GRCh38

View Full Product Details