Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244518587-244518812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf101
Entrez Gene ID
257044
Gene Name
chromosome 1 open reading frame 101
Gene Aliases
-
UniGene
Hs.459534
Gene Chromosome Location
Chr.1: 244452014-244640362 on Build GRCh38
Transcripts
NM_001130957.1 NM_001242340.1 NM_173807.4 XM_011544138.2 XM_011544139.2 XM_011544142.2 XM_011544144.2 XM_011544145.2 XM_011544149.2 XM_011544151.2 XM_017000943.1 XM_017000944.1 XM_017000945.1 XM_017000946.1 XM_017000947.1 XM_017000948.1 XM_017000949.1 XM_017000950.1 XM_017000951.1 XM_017000952.1 XM_017000953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
425805 c.512C>G p.S171C Chr.1: 244518674-244518674 on Build GRCh38
1340801 c.522G>T p.E174D Chr.1: 244518684-244518684 on Build GRCh38
1473721 c.512C>G p.S171C Chr.1: 244518674-244518674 on Build GRCh38
1340800 c.522G>T p.E174D Chr.1: 244518684-244518684 on Build GRCh38
7291273 c.446G>C p.S149T Chr.1: 244518608-244518608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769577999 Chr.1: 244518711-244518711 on Build GRCh38
rs780586666 Chr.1: 244518712-244518713 on Build GRCh38
rs1048113157 Chr.1: 244518753-244518753 on Build GRCh38
rs115284937 Chr.1: 244518774-244518774 on Build GRCh38
rs746074925 Chr.1: 244518696-244518696 on Build GRCh38
rs767294747 Chr.1: 244518719-244518719 on Build GRCh38
rs778361586 Chr.1: 244518653-244518653 on Build GRCh38
rs763740783 Chr.1: 244518663-244518663 on Build GRCh38
rs763034099 Chr.1: 244518715-244518715 on Build GRCh38
rs755045783 Chr.1: 244518723-244518724 on Build GRCh38
rs765976206 Chr.1: 244518716-244518716 on Build GRCh38
rs761128393 Chr.1: 244518749-244518749 on Build GRCh38
rs752959547 Chr.1: 244518746-244518746 on Build GRCh38
rs748553799 Chr.1: 244518623-244518623 on Build GRCh38
rs750733816 Chr.1: 244518707-244518707 on Build GRCh38
rs772548361 Chr.1: 244518622-244518622 on Build GRCh38
rs368338238 Chr.1: 244518637-244518637 on Build GRCh38
rs753113480 Chr.1: 244518650-244518650 on Build GRCh38
rs572782580 Chr.1: 244518628-244518628 on Build GRCh38
rs555702665 Chr.1: 244518718-244518718 on Build GRCh38
rs147923178 Chr.1: 244518684-244518684 on Build GRCh38
rs764996062 Chr.1: 244518706-244518707 on Build GRCh38
rs896910384 Chr.1: 244518714-244518714 on Build GRCh38
rs753540943 Chr.1: 244518678-244518685 on Build GRCh38
rs767992327 Chr.1: 244518647-244518647 on Build GRCh38
rs867910076 Chr.1: 244518693-244518694 on Build GRCh38
rs1051852516 Chr.1: 244518763-244518763 on Build GRCh38
rs751236170 Chr.1: 244518629-244518629 on Build GRCh38
rs189730930 Chr.1: 244518697-244518697 on Build GRCh38
rs757293745 Chr.1: 244518669-244518669 on Build GRCh38
rs781306413 Chr.1: 244518725-244518726 on Build GRCh38
rs756525241 Chr.1: 244518651-244518651 on Build GRCh38
rs760243738 Chr.1: 244518657-244518661 on Build GRCh38
rs752045676 Chr.1: 244518719-244518720 on Build GRCh38
rs749284761 Chr.1: 244518660-244518660 on Build GRCh38
rs888106959 Chr.1: 244518758-244518758 on Build GRCh38
rs142019600 Chr.1: 244518671-244518671 on Build GRCh38
rs776221312 Chr.1: 244518699-244518699 on Build GRCh38
rs947678067 Chr.1: 244518655-244518655 on Build GRCh38
rs765224753 Chr.1: 244518614-244518614 on Build GRCh38
rs544634919 Chr.1: 244518769-244518769 on Build GRCh38
rs773171499 Chr.1: 244518713-244518713 on Build GRCh38
rs757029022 Chr.1: 244518694-244518695 on Build GRCh38
rs758723396 Chr.1: 244518709-244518710 on Build GRCh38

View Full Product Details