Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96012629-96012854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D2B
Entrez Gene ID
387707
Gene Name
coiled-coil and C2 domain containing 2B
Gene Aliases
C10orf130, bA248J23.4
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95974029-96035167 on Build GRCh38
Transcripts
NM_001001732.3 NM_001159747.1 XM_011539789.2 XM_011539790.2 XM_011539791.2 XM_011539792.2 XM_011539793.2 XM_011539795.2 XM_017016240.1 NR_038444.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001001732.3 NM_001159747.1 XM_011539789.2 XM_011539790.2 XM_011539791.2 XM_011539792.2 XM_011539793.2 XM_011539795.2 XM_017016240.1 NR_038444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4537439 c.241G>A p.V81I Chr.10: 96012652-96012652 on Build GRCh38
4537438 c.241G>A p.V81I Chr.10: 96012652-96012652 on Build GRCh38
7101967 c.290T>C p.I97T Chr.10: 96012701-96012701 on Build GRCh38
1350056 c.282T>A p.L94L Chr.10: 96012693-96012693 on Build GRCh38
4565603 c.227_228GG>AA p.G76E Chr.10: 96012638-96012639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756682422 Chr.10: 96012685-96012685 on Build GRCh38
rs781685427 Chr.10: 96012711-96012711 on Build GRCh38
rs746015743 Chr.10: 96012778-96012779 on Build GRCh38
rs753536602 Chr.10: 96012673-96012673 on Build GRCh38
rs887457359 Chr.10: 96012798-96012798 on Build GRCh38
rs10748652 Chr.10: 96012739-96012739 on Build GRCh38
rs778508451 Chr.10: 96012703-96012703 on Build GRCh38
rs1048676619 Chr.10: 96012764-96012764 on Build GRCh38
rs770864858 Chr.10: 96012772-96012772 on Build GRCh38
rs747498895 Chr.10: 96012706-96012706 on Build GRCh38
rs918838366 Chr.10: 96012713-96012713 on Build GRCh38
rs375134007 Chr.10: 96012664-96012664 on Build GRCh38
rs867604733 Chr.10: 96012671-96012671 on Build GRCh38
rs763979866 Chr.10: 96012781-96012782 on Build GRCh38
rs755752801 Chr.10: 96012661-96012661 on Build GRCh38
rs560336535 Chr.10: 96012730-96012730 on Build GRCh38
rs773236724 Chr.10: 96012754-96012754 on Build GRCh38
rs769324599 Chr.10: 96012710-96012710 on Build GRCh38
rs763262515 Chr.10: 96012767-96012767 on Build GRCh38
rs949572421 Chr.10: 96012660-96012660 on Build GRCh38
rs775831674 Chr.10: 96012780-96012781 on Build GRCh38
rs943111058 Chr.10: 96012810-96012810 on Build GRCh38

View Full Product Details