Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60429315-60429586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf64
Entrez Gene ID
124773
Gene Name
chromosome 17 open reading frame 64
Gene Aliases
-
UniGene
Hs.129312
Gene Chromosome Location
Chr.17: 60421091-60431426 on Build GRCh38
Transcripts
NM_181707.2 XM_005257033.4 XM_005257034.4 XM_005257035.4 XM_005257036.2 XM_017024166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1589098 c.598C>T p.P200S Chr.17: 60429530-60429530 on Build GRCh38
4549721 c.480G>A p.R160R Chr.17: 60429412-60429412 on Build GRCh38
1589097 c.603A>G p.R201R Chr.17: 60429535-60429535 on Build GRCh38
1610562 c.447G>A p.S149S Chr.17: 60429379-60429379 on Build GRCh38
5972325 c.127G>A p.E43K Chr.17: 60429389-60429389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757380106 Chr.17: 60429485-60429485 on Build GRCh38
rs777994062 Chr.17: 60429355-60429355 on Build GRCh38
rs201950282 Chr.17: 60429557-60429557 on Build GRCh38
rs751251927 Chr.17: 60429494-60429494 on Build GRCh38
rs201450863 Chr.17: 60429407-60429407 on Build GRCh38
rs756290785 Chr.17: 60429354-60429354 on Build GRCh38
rs200569085 Chr.17: 60429385-60429385 on Build GRCh38
rs140000868 Chr.17: 60429393-60429393 on Build GRCh38
rs367735553 Chr.17: 60429466-60429466 on Build GRCh38
rs76238897 Chr.17: 60429566-60429566 on Build GRCh38
rs1045172010 Chr.17: 60429436-60429436 on Build GRCh38
rs200962057 Chr.17: 60429437-60429437 on Build GRCh38
rs747591049 Chr.17: 60429416-60429416 on Build GRCh38
rs905169331 Chr.17: 60429350-60429350 on Build GRCh38
rs760760620 Chr.17: 60429454-60429454 on Build GRCh38
rs763620676 Chr.17: 60429442-60429443 on Build GRCh38
rs944359779 Chr.17: 60429417-60429417 on Build GRCh38
rs1027739290 Chr.17: 60429490-60429490 on Build GRCh38
rs9913666 Chr.17: 60429424-60429424 on Build GRCh38
rs910549952 Chr.17: 60429367-60429367 on Build GRCh38
rs777905878 Chr.17: 60429397-60429397 on Build GRCh38
rs1056158164 Chr.17: 60429477-60429477 on Build GRCh38
rs141815013 Chr.17: 60429373-60429373 on Build GRCh38
rs199998109 Chr.17: 60429388-60429388 on Build GRCh38
rs775980827 Chr.17: 60429470-60429470 on Build GRCh38
rs371681055 Chr.17: 60429500-60429500 on Build GRCh38
rs754046740 Chr.17: 60429400-60429400 on Build GRCh38
rs756200782 Chr.17: 60429396-60429396 on Build GRCh38
rs539594178 Chr.17: 60429513-60429513 on Build GRCh38
rs746897514 Chr.17: 60429370-60429370 on Build GRCh38
rs753989446 Chr.17: 60429481-60429481 on Build GRCh38
rs763428300 Chr.17: 60429339-60429339 on Build GRCh38
rs370364055 Chr.17: 60429455-60429455 on Build GRCh38
rs368635684 Chr.17: 60429467-60429467 on Build GRCh38
rs748873934 Chr.17: 60429442-60429442 on Build GRCh38
rs761228169 Chr.17: 60429474-60429474 on Build GRCh38
rs762738412 Chr.17: 60429379-60429379 on Build GRCh38
rs781107492 Chr.17: 60429353-60429353 on Build GRCh38
rs201288791 Chr.17: 60429361-60429361 on Build GRCh38
rs143631455 Chr.17: 60429401-60429401 on Build GRCh38
rs755563587 Chr.17: 60429347-60429347 on Build GRCh38
rs142915363 Chr.17: 60429487-60429487 on Build GRCh38
rs752725736 Chr.17: 60429389-60429389 on Build GRCh38
rs769510201 Chr.17: 60429374-60429374 on Build GRCh38
rs779956263 Chr.17: 60429363-60429363 on Build GRCh38
rs374619858 Chr.17: 60429412-60429412 on Build GRCh38
rs952436367 Chr.17: 60429526-60429526 on Build GRCh38
rs768426490 Chr.17: 60429411-60429411 on Build GRCh38
rs756172965 Chr.17: 60429535-60429535 on Build GRCh38
rs777390297 Chr.17: 60429547-60429547 on Build GRCh38
rs766847422 Chr.17: 60429346-60429346 on Build GRCh38
rs574216649 Chr.17: 60429447-60429447 on Build GRCh38
rs61747124 Chr.17: 60429406-60429406 on Build GRCh38
rs1045322464 Chr.17: 60429558-60429558 on Build GRCh38
rs760613774 Chr.17: 60429335-60429336 on Build GRCh38
rs748089677 Chr.17: 60429514-60429514 on Build GRCh38
rs770482041 Chr.17: 60429345-60429345 on Build GRCh38
rs1003725055 Chr.17: 60429476-60429476 on Build GRCh38
rs1007801819 Chr.17: 60429527-60429527 on Build GRCh38
rs768677332 Chr.17: 60429371-60429371 on Build GRCh38
rs905221901 Chr.17: 60429562-60429562 on Build GRCh38
rs771760090 Chr.17: 60429362-60429362 on Build GRCh38
rs57928695 Chr.17: 60429387-60429387 on Build GRCh38
rs572279125 Chr.17: 60429480-60429480 on Build GRCh38

View Full Product Details