Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81061794-81062039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf46
Entrez Gene ID
123775
Gene Name
chromosome 16 open reading frame 46
Gene Aliases
-
UniGene
Hs.658684
Gene Chromosome Location
Chr.16: 81053497-81077267 on Build GRCh38
Transcripts
NM_001100873.1 NM_152337.2 XM_005255792.2 XM_011522848.2 XM_011522849.2 XM_017022916.1 XM_017022917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5690296 c.362G>T p.G121V Chr.16: 81061987-81061987 on Build GRCh38
5580395 c.431C>T p.S144F Chr.16: 81061918-81061918 on Build GRCh38
4489210 c.344C>A p.T115N Chr.16: 81062005-81062005 on Build GRCh38
1588556 c.469C>T p.R157* Chr.16: 81061880-81061880 on Build GRCh38
5363783 c.453C>G p.C151W Chr.16: 81061896-81061896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967588114 Chr.16: 81061831-81061831 on Build GRCh38
rs779033308 Chr.16: 81061830-81061830 on Build GRCh38
rs780125351 Chr.16: 81061904-81061904 on Build GRCh38
rs906361568 Chr.16: 81061986-81061986 on Build GRCh38
rs757854301 Chr.16: 81061842-81061842 on Build GRCh38
rs761730810 Chr.16: 81061814-81061814 on Build GRCh38
rs137925772 Chr.16: 81061823-81061823 on Build GRCh38
rs766163636 Chr.16: 81061978-81061978 on Build GRCh38
rs1002980905 Chr.16: 81062007-81062007 on Build GRCh38
rs760426393 Chr.16: 81061890-81061890 on Build GRCh38
rs35597223 Chr.16: 81061869-81061869 on Build GRCh38
rs939935320 Chr.16: 81061961-81061961 on Build GRCh38
rs747020676 Chr.16: 81061839-81061839 on Build GRCh38
rs150847199 Chr.16: 81061930-81061930 on Build GRCh38
rs552328735 Chr.16: 81061931-81061931 on Build GRCh38
rs529021063 Chr.16: 81061822-81061822 on Build GRCh38
rs149630546 Chr.16: 81061833-81061833 on Build GRCh38
rs990702647 Chr.16: 81061901-81061901 on Build GRCh38
rs975555889 Chr.16: 81061879-81061879 on Build GRCh38
rs777995234 Chr.16: 81061834-81061834 on Build GRCh38
rs145595395 Chr.16: 81061880-81061880 on Build GRCh38
rs780163711 Chr.16: 81061825-81061826 on Build GRCh38
rs764444536 Chr.16: 81061992-81061992 on Build GRCh38
rs773943085 Chr.16: 81061935-81061935 on Build GRCh38
rs766185049 Chr.16: 81061902-81061902 on Build GRCh38
rs773853282 Chr.16: 81061871-81061871 on Build GRCh38
rs758700250 Chr.16: 81061908-81061908 on Build GRCh38
rs747685632 Chr.16: 81061949-81061949 on Build GRCh38
rs376500530 Chr.16: 81061912-81061912 on Build GRCh38
rs753998253 Chr.16: 81061937-81061937 on Build GRCh38
rs756490402 Chr.16: 81061928-81061928 on Build GRCh38
rs535125163 Chr.16: 81061981-81061981 on Build GRCh38
rs759358077 Chr.16: 81061984-81061984 on Build GRCh38
rs533239952 Chr.16: 81061888-81061888 on Build GRCh38
rs781747558 Chr.16: 81061851-81061851 on Build GRCh38
rs767463649 Chr.16: 81061884-81061884 on Build GRCh38
rs908476644 Chr.16: 81061954-81061954 on Build GRCh38
rs773071105 Chr.16: 81061957-81061957 on Build GRCh38
rs144394384 Chr.16: 81061866-81061866 on Build GRCh38
rs756424839 Chr.16: 81061861-81061861 on Build GRCh38
rs930992894 Chr.16: 81061905-81061905 on Build GRCh38
rs574719697 Chr.16: 81061982-81061982 on Build GRCh38
rs200088357 Chr.16: 81061988-81061988 on Build GRCh38
rs748952913 Chr.16: 81061868-81061868 on Build GRCh38
rs757375444 Chr.16: 81061918-81061918 on Build GRCh38
rs746367231 Chr.16: 81061924-81061924 on Build GRCh38
rs781594740 Chr.16: 81061922-81061922 on Build GRCh38
rs751802794 Chr.16: 81061993-81061993 on Build GRCh38
rs764356197 Chr.16: 81061910-81061910 on Build GRCh38
rs757465481 Chr.16: 81061997-81061997 on Build GRCh38
rs139640227 Chr.16: 81061956-81061956 on Build GRCh38
rs770168376 Chr.16: 81061864-81061864 on Build GRCh38
rs200288207 Chr.16: 81061816-81061816 on Build GRCh38
rs377690413 Chr.16: 81061965-81061965 on Build GRCh38
rs767371983 Chr.16: 81061815-81061815 on Build GRCh38
rs989531596 Chr.16: 81062006-81062006 on Build GRCh38
rs917739173 Chr.16: 81061920-81061920 on Build GRCh38

View Full Product Details