Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 88139733-88139978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGGBP1
Entrez Gene ID
8545
Gene Name
CGG triplet repeat binding protein 1
Gene Aliases
CGGBP, p20-CGGBP
UniGene
Hs.444818
Gene Chromosome Location
Chr.3: 88051950-88149866 on Build GRCh38
Transcripts
NM_001195308.1 NM_018293.2 XM_011534340.2 XM_017006789.1 XM_017006790.1 XM_017006791.1 XM_017006792.1 XM_017006793.1 XM_017006794.1 XM_017006795.1

Gene Symbol
ZNF654
Entrez Gene ID
55279
Gene Name
zinc finger protein 654
Gene Aliases
-
UniGene
Hs.591650
Gene Chromosome Location
Chr.3: 88059217-88144664 on Build GRCh38
Transcripts
NM_001195308.1 NM_018293.2 XM_011534340.2 XM_017006789.1 XM_017006790.1 XM_017006791.1 XM_017006792.1 XM_017006793.1 XM_017006794.1 XM_017006795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1741629 c.484G>A p.D162N Chr.3: 88139794-88139794 on Build GRCh38
7203663 c.542A>G p.N181S Chr.3: 88139852-88139852 on Build GRCh38
4120468 c.505C>T p.R169W Chr.3: 88139932-88139932 on Build GRCh38
4120467 c.622C>T p.R208W Chr.3: 88139932-88139932 on Build GRCh38
6832124 c.506G>A p.R169Q Chr.3: 88139933-88139933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766111404 Chr.3: 88139860-88139860 on Build GRCh38
rs759026152 Chr.3: 88139932-88139932 on Build GRCh38
rs753231217 Chr.3: 88139861-88139861 on Build GRCh38
rs368811288 Chr.3: 88139863-88139863 on Build GRCh38
rs375388602 Chr.3: 88139852-88139852 on Build GRCh38
rs749848210 Chr.3: 88139762-88139767 on Build GRCh38
rs536331654 Chr.3: 88139909-88139909 on Build GRCh38
rs767863222 Chr.3: 88139818-88139818 on Build GRCh38
rs983170470 Chr.3: 88139933-88139933 on Build GRCh38
rs952381399 Chr.3: 88139859-88139859 on Build GRCh38
rs568647405 Chr.3: 88139796-88139796 on Build GRCh38
rs773520518 Chr.3: 88139805-88139805 on Build GRCh38
rs771283678 Chr.3: 88139800-88139802 on Build GRCh38
rs748688667 Chr.3: 88139922-88139922 on Build GRCh38
rs975420426 Chr.3: 88139850-88139850 on Build GRCh38
rs754521302 Chr.3: 88139874-88139874 on Build GRCh38
rs755715010 Chr.3: 88139858-88139858 on Build GRCh38
rs778253782 Chr.3: 88139950-88139950 on Build GRCh38
rs878897642 Chr.3: 88139941-88139941 on Build GRCh38
rs761543001 Chr.3: 88139851-88139851 on Build GRCh38
rs763868413 Chr.3: 88139836-88139836 on Build GRCh38
rs371749343 Chr.3: 88139918-88139918 on Build GRCh38
rs747216698 Chr.3: 88139814-88139814 on Build GRCh38
rs1019080178 Chr.3: 88139783-88139783 on Build GRCh38
rs772595992 Chr.3: 88139784-88139784 on Build GRCh38
rs569469594 Chr.3: 88139832-88139832 on Build GRCh38
rs537595025 Chr.3: 88139840-88139840 on Build GRCh38
rs750258005 Chr.3: 88139853-88139853 on Build GRCh38
rs774298577 Chr.3: 88139848-88139848 on Build GRCh38
rs776571548 Chr.3: 88139823-88139823 on Build GRCh38

View Full Product Details