Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134918726-134918995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNT2
Entrez Gene ID
905
Gene Name
cyclin T2
Gene Aliases
CYCT2
UniGene
Hs.744115
Gene Chromosome Location
Chr.2: 134918810-134959345 on Build GRCh38
Transcripts
NM_001241.3 NM_001320748.1 NM_001320749.1 NM_058241.2 NR_037649.1 NR_135468.1 XM_017005227.1 XM_017005228.1 XM_017005229.1 XM_017005230.1 XM_017005231.1 XM_017005232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5489864 c.115C>T p.Q39* Chr.2: 134918969-134918969 on Build GRCh38
3566659 c.54G>A p.L18L Chr.2: 134918908-134918908 on Build GRCh38
6209060 c.90G>T p.A30A Chr.2: 134918944-134918944 on Build GRCh38
6623110 c.20_22delCTT p.S9delS Chr.2: 134918874-134918876 on Build GRCh38
1200182 c.134A>G p.Q45R Chr.2: 134918988-134918988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546616266 Chr.2: 134918753-134918753 on Build GRCh38
rs748908380 Chr.2: 134918843-134918843 on Build GRCh38
rs777316708 Chr.2: 134918913-134918913 on Build GRCh38
rs780933546 Chr.2: 134918884-134918884 on Build GRCh38
rs777242207 Chr.2: 134918842-134918842 on Build GRCh38
rs1053707433 Chr.2: 134918827-134918827 on Build GRCh38
rs922072674 Chr.2: 134918792-134918792 on Build GRCh38
rs528679108 Chr.2: 134918911-134918911 on Build GRCh38
rs774737511 Chr.2: 134918816-134918816 on Build GRCh38
rs772512537 Chr.2: 134918937-134918937 on Build GRCh38
rs754116930 Chr.2: 134918823-134918823 on Build GRCh38
rs752718185 Chr.2: 134918819-134918819 on Build GRCh38
rs1002715560 Chr.2: 134918870-134918870 on Build GRCh38
rs987927057 Chr.2: 134918803-134918803 on Build GRCh38
rs1044239920 Chr.2: 134918902-134918902 on Build GRCh38
rs750316661 Chr.2: 134918816-134918817 on Build GRCh38
rs758280660 Chr.2: 134918817-134918817 on Build GRCh38
rs1033693097 Chr.2: 134918974-134918974 on Build GRCh38
rs779959916 Chr.2: 134918831-134918831 on Build GRCh38
rs954916483 Chr.2: 134918795-134918795 on Build GRCh38
rs557003975 Chr.2: 134918818-134918818 on Build GRCh38
rs1008131661 Chr.2: 134918768-134918768 on Build GRCh38
rs770619354 Chr.2: 134918849-134918849 on Build GRCh38
rs968488344 Chr.2: 134918774-134918774 on Build GRCh38
rs746177381 Chr.2: 134918928-134918928 on Build GRCh38
rs945474574 Chr.2: 134918894-134918894 on Build GRCh38
rs879520100 Chr.2: 134918751-134918751 on Build GRCh38
rs146432822 Chr.2: 134918877-134918877 on Build GRCh38
rs1029256280 Chr.2: 134918954-134918954 on Build GRCh38
rs956560176 Chr.2: 134918802-134918802 on Build GRCh38
rs774686197 Chr.2: 134918850-134918850 on Build GRCh38
rs760816765 Chr.2: 134918822-134918822 on Build GRCh38
rs547635001 Chr.2: 134918787-134918787 on Build GRCh38
rs1004552613 Chr.2: 134918763-134918763 on Build GRCh38
rs1022282174 Chr.2: 134918762-134918762 on Build GRCh38
rs751446411 Chr.2: 134918832-134918832 on Build GRCh38
rs568358031 Chr.2: 134918796-134918796 on Build GRCh38
rs772395329 Chr.2: 134918863-134918863 on Build GRCh38
rs528271737 Chr.2: 134918748-134918748 on Build GRCh38
rs1002580391 Chr.2: 134918914-134918914 on Build GRCh38
rs765165323 Chr.2: 134918814-134918829 on Build GRCh38
rs377346816 Chr.2: 134918879-134918879 on Build GRCh38
rs770568165 Chr.2: 134918923-134918923 on Build GRCh38
rs996403155 Chr.2: 134918942-134918942 on Build GRCh38
rs1015975496 Chr.2: 134918766-134918766 on Build GRCh38
rs769088871 Chr.2: 134918959-134918959 on Build GRCh38
rs752702499 Chr.2: 134918897-134918897 on Build GRCh38
rs976487079 Chr.2: 134918791-134918791 on Build GRCh38
rs760626499 Chr.2: 134918868-134918868 on Build GRCh38
rs766762498 Chr.2: 134918814-134918814 on Build GRCh38
rs762102247 Chr.2: 134918878-134918878 on Build GRCh38
rs758222548 Chr.2: 134918824-134918824 on Build GRCh38
rs1019078306 Chr.2: 134918773-134918773 on Build GRCh38
rs751393482 Chr.2: 134918882-134918882 on Build GRCh38
rs905662918 Chr.2: 134918862-134918862 on Build GRCh38
rs575574074 Chr.2: 134918944-134918944 on Build GRCh38
rs200294959 Chr.2: 134918835-134918835 on Build GRCh38
rs370795827 Chr.2: 134918866-134918866 on Build GRCh38
rs776835071 Chr.2: 134918962-134918962 on Build GRCh38
rs764269176 Chr.2: 134918871-134918871 on Build GRCh38
rs778544283 Chr.2: 134918926-134918926 on Build GRCh38
rs570579166 Chr.2: 134918745-134918745 on Build GRCh38
rs1002458556 Chr.2: 134918785-134918785 on Build GRCh38
rs779683041 Chr.2: 134918898-134918898 on Build GRCh38
rs760015988 Chr.2: 134918817-134918817 on Build GRCh38
rs577406279 Chr.2: 134918917-134918917 on Build GRCh38
rs775824439 Chr.2: 134918941-134918941 on Build GRCh38
rs535699679 Chr.2: 134918798-134918798 on Build GRCh38
rs781255667 Chr.2: 134918836-134918836 on Build GRCh38
rs759962696 Chr.2: 134918854-134918854 on Build GRCh38
rs1015921708 Chr.2: 134918887-134918887 on Build GRCh38
rs761209997 Chr.2: 134918947-134918947 on Build GRCh38
rs753899934 Chr.2: 134918876-134918876 on Build GRCh38
rs975018347 Chr.2: 134918789-134918789 on Build GRCh38
rs1010090384 Chr.2: 134918757-134918757 on Build GRCh38
rs965058950 Chr.2: 134918778-134918778 on Build GRCh38
rs45611231 Chr.2: 134918839-134918839 on Build GRCh38

View Full Product Details