Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31640606-31640879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
UniGene
Hs.440900
Gene Chromosome Location
Chr.6: 31639028-31660900 on Build GRCh38
Transcripts
NM_001098534.1 NM_001199697.1 NM_001199698.1 NM_004639.3 NM_080702.2 NM_080703.2 XM_011514892.2 XM_017011275.1 XM_017011276.1 XM_017011277.1 XM_017011278.1 XM_017011279.1 XM_017011280.1 XM_017011281.1 XM_017011282.1 XM_017011283.1 XM_017011284.1 XM_017011285.1 XM_017011286.1 XM_017011287.1 XM_017011288.1 XM_017011289.1 XM_017011290.1 XM_017011291.1 XM_017011292.1 XM_017011293.1 XM_017011294.1 XM_017011295.1 XM_017011296.1 XM_017011297.1 XM_017011298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6024295 c.2823_2824insC p.Q942fs*5 Chr.6: 31640794-31640795 on Build GRCh38
4786876 c.2953A>G p.M985V Chr.6: 31640686-31640686 on Build GRCh38
3624642 c.2895C>T p.A965A Chr.6: 31640831-31640831 on Build GRCh38
1443307 c.2845A>G p.M949V Chr.6: 31640686-31640686 on Build GRCh38
3624641 c.2787C>T p.A929A Chr.6: 31640831-31640831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757208756 Chr.6: 31640852-31640852 on Build GRCh38
rs769446010 Chr.6: 31640741-31640741 on Build GRCh38
rs968405450 Chr.6: 31640706-31640706 on Build GRCh38
rs776415085 Chr.6: 31640654-31640654 on Build GRCh38
rs138429242 Chr.6: 31640819-31640819 on Build GRCh38
rs760904441 Chr.6: 31640698-31640698 on Build GRCh38
rs368745016 Chr.6: 31640773-31640773 on Build GRCh38
rs777860050 Chr.6: 31640631-31640631 on Build GRCh38
rs373154403 Chr.6: 31640746-31640746 on Build GRCh38
rs138969324 Chr.6: 31640818-31640818 on Build GRCh38
rs781178827 Chr.6: 31640726-31640726 on Build GRCh38
rs773588977 Chr.6: 31640810-31640810 on Build GRCh38
rs757083601 Chr.6: 31640723-31640723 on Build GRCh38
rs748773115 Chr.6: 31640761-31640761 on Build GRCh38
rs781019721 Chr.6: 31640858-31640858 on Build GRCh38
rs754070310 Chr.6: 31640825-31640825 on Build GRCh38
rs903697326 Chr.6: 31640679-31640679 on Build GRCh38
rs750589900 Chr.6: 31640697-31640697 on Build GRCh38
rs765443408 Chr.6: 31640832-31640832 on Build GRCh38
rs1044398958 Chr.6: 31640786-31640786 on Build GRCh38
rs766542591 Chr.6: 31640699-31640699 on Build GRCh38
rs144562066 Chr.6: 31640830-31640830 on Build GRCh38
rs554159674 Chr.6: 31640796-31640796 on Build GRCh38
rs746746906 Chr.6: 31640632-31640632 on Build GRCh38
rs377158172 Chr.6: 31640776-31640776 on Build GRCh38
rs745669719 Chr.6: 31640729-31640729 on Build GRCh38
rs777843651 Chr.6: 31640708-31640708 on Build GRCh38
rs150422210 Chr.6: 31640686-31640686 on Build GRCh38
rs778695310 Chr.6: 31640793-31640794 on Build GRCh38
rs751517791 Chr.6: 31640838-31640838 on Build GRCh38
rs767909809 Chr.6: 31640688-31640688 on Build GRCh38
rs762152262 Chr.6: 31640687-31640687 on Build GRCh38
rs751618508 Chr.6: 31640709-31640709 on Build GRCh38
rs769162196 Chr.6: 31640665-31640665 on Build GRCh38
rs369392276 Chr.6: 31640701-31640701 on Build GRCh38
rs534951276 Chr.6: 31640641-31640641 on Build GRCh38
rs762398479 Chr.6: 31640680-31640680 on Build GRCh38
rs745570214 Chr.6: 31640656-31640656 on Build GRCh38
rs950007001 Chr.6: 31640738-31640738 on Build GRCh38
rs111994917 Chr.6: 31640644-31640644 on Build GRCh38
rs34061730 Chr.6: 31640702-31640702 on Build GRCh38
rs775013758 Chr.6: 31640672-31640672 on Build GRCh38

View Full Product Details