Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38871475-38871742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX6
Entrez Gene ID
23466
Gene Name
chromobox 6
Gene Aliases
-
UniGene
Hs.592201
Gene Chromosome Location
Chr.22: 38861427-38872334 on Build GRCh38
Transcripts
NM_001303494.1 NM_014292.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7105673 c.202G>C p.G68R Chr.22: 38871524-38871524 on Build GRCh38
6282160 c.181G>A p.E61K Chr.22: 38871545-38871545 on Build GRCh38
3363646 c.146T>A p.L49Q Chr.22: 38871725-38871725 on Build GRCh38
5941072 c.238C>T p.L80F Chr.22: 38871488-38871488 on Build GRCh38
3424178 c.210G>T p.K70N Chr.22: 38871516-38871516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025276124 Chr.22: 38871604-38871604 on Build GRCh38
rs374680103 Chr.22: 38871721-38871721 on Build GRCh38
rs201405992 Chr.22: 38871683-38871683 on Build GRCh38
rs780009114 Chr.22: 38871588-38871588 on Build GRCh38
rs768026232 Chr.22: 38871703-38871703 on Build GRCh38
rs367836108 Chr.22: 38871536-38871536 on Build GRCh38
rs777554115 Chr.22: 38871681-38871683 on Build GRCh38
rs540910085 Chr.22: 38871563-38871563 on Build GRCh38
rs758315779 Chr.22: 38871644-38871645 on Build GRCh38
rs138074053 Chr.22: 38871665-38871665 on Build GRCh38
rs776056339 Chr.22: 38871709-38871709 on Build GRCh38
rs748238959 Chr.22: 38871658-38871658 on Build GRCh38
rs553600038 Chr.22: 38871583-38871583 on Build GRCh38
rs771241347 Chr.22: 38871671-38871671 on Build GRCh38
rs752695107 Chr.22: 38871581-38871606 on Build GRCh38
rs774771314 Chr.22: 38871559-38871559 on Build GRCh38
rs74361001 Chr.22: 38871569-38871569 on Build GRCh38
rs759913032 Chr.22: 38871678-38871678 on Build GRCh38
rs770132342 Chr.22: 38871663-38871663 on Build GRCh38
rs145539395 Chr.22: 38871652-38871652 on Build GRCh38
rs763474622 Chr.22: 38871568-38871568 on Build GRCh38
rs868594192 Chr.22: 38871538-38871538 on Build GRCh38
rs776880966 Chr.22: 38871509-38871509 on Build GRCh38
rs766894303 Chr.22: 38871558-38871558 on Build GRCh38
rs3885298 Chr.22: 38871635-38871635 on Build GRCh38
rs773623052 Chr.22: 38871664-38871664 on Build GRCh38
rs200227732 Chr.22: 38871551-38871551 on Build GRCh38
rs1027164055 Chr.22: 38871602-38871602 on Build GRCh38
rs375896091 Chr.22: 38871648-38871648 on Build GRCh38
rs768657282 Chr.22: 38871508-38871508 on Build GRCh38
rs199673563 Chr.22: 38871579-38871579 on Build GRCh38
rs139570238 Chr.22: 38871510-38871510 on Build GRCh38
rs1029263549 Chr.22: 38871627-38871627 on Build GRCh38
rs764875072 Chr.22: 38871578-38871578 on Build GRCh38
rs565167495 Chr.22: 38871597-38871597 on Build GRCh38
rs532525908 Chr.22: 38871625-38871625 on Build GRCh38
rs746879261 Chr.22: 38871591-38871591 on Build GRCh38
rs369373813 Chr.22: 38871557-38871557 on Build GRCh38
rs758186283 Chr.22: 38871587-38871587 on Build GRCh38
rs774811478 Chr.22: 38871674-38871674 on Build GRCh38

View Full Product Details