Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30834532-30834757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD5
Entrez Gene ID
79915
Gene Name
ATPase family, AAA domain containing 5
Gene Aliases
C17orf41, ELG1, FRAG1
UniGene
Hs.528902
Gene Chromosome Location
Chr.17: 30830932-30896424 on Build GRCh38
Transcripts
NM_024857.4 XM_011525269.2 XM_011525270.2 XM_011525273.2 XM_017025111.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889744 c.551C>T p.S184F Chr.17: 30834632-30834632 on Build GRCh38
332698 c.470C>T p.S157F Chr.17: 30834551-30834551 on Build GRCh38
6875770 c.663C>T p.P221P Chr.17: 30834744-30834744 on Build GRCh38
6606429 c.486G>T p.K162N Chr.17: 30834567-30834567 on Build GRCh38
7108257 c.632G>C p.R211T Chr.17: 30834713-30834713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768574370 Chr.17: 30834639-30834639 on Build GRCh38
rs914996435 Chr.17: 30834629-30834629 on Build GRCh38
rs759319232 Chr.17: 30834671-30834671 on Build GRCh38
rs181925781 Chr.17: 30834573-30834573 on Build GRCh38
rs773461644 Chr.17: 30834672-30834672 on Build GRCh38
rs774244442 Chr.17: 30834662-30834662 on Build GRCh38
rs760661424 Chr.17: 30834677-30834677 on Build GRCh38
rs367699401 Chr.17: 30834724-30834724 on Build GRCh38
rs372570681 Chr.17: 30834720-30834720 on Build GRCh38
rs745340855 Chr.17: 30834620-30834620 on Build GRCh38
rs1042030653 Chr.17: 30834694-30834694 on Build GRCh38
rs775276466 Chr.17: 30834624-30834624 on Build GRCh38
rs746696655 Chr.17: 30834568-30834568 on Build GRCh38
rs749190277 Chr.17: 30834632-30834632 on Build GRCh38
rs150438654 Chr.17: 30834729-30834729 on Build GRCh38
rs1047706402 Chr.17: 30834606-30834606 on Build GRCh38
rs762145513 Chr.17: 30834663-30834663 on Build GRCh38
rs1018482018 Chr.17: 30834652-30834652 on Build GRCh38
rs755474549 Chr.17: 30834701-30834701 on Build GRCh38
rs780816463 Chr.17: 30834732-30834732 on Build GRCh38
rs766428559 Chr.17: 30834680-30834680 on Build GRCh38
rs780730225 Chr.17: 30834588-30834588 on Build GRCh38
rs769837150 Chr.17: 30834623-30834623 on Build GRCh38

View Full Product Details