Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 53967019-53967244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS1
Entrez Gene ID
8537
Gene Name
breast carcinoma amplified sequence 1
Gene Aliases
AIBC1, NABC1
UniGene
Hs.400556
Gene Chromosome Location
Chr.20: 53943538-54070765 on Build GRCh38
Transcripts
NM_001316361.2 NM_001323347.1 NM_003657.3 XM_005260587.2 XM_005260589.2 XM_005260590.2 XM_005260591.2 XM_005260593.2 XM_005260594.2 XM_005260595.2 XM_011529090.2 XM_011529091.2 XM_017028110.1 XM_017028111.1 XM_017028112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4099579 c.1320A>T p.V440V Chr.20: 53967029-53967029 on Build GRCh38
6869724 c.1208T>A p.I403N Chr.20: 53967048-53967048 on Build GRCh38
1283639 c.1217C>T p.S406F Chr.20: 53967039-53967039 on Build GRCh38
4099578 c.1227A>T p.V409V Chr.20: 53967029-53967029 on Build GRCh38
6869725 c.1301T>A p.I434N Chr.20: 53967048-53967048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759265798 Chr.20: 53967055-53967055 on Build GRCh38
rs532261400 Chr.20: 53967197-53967197 on Build GRCh38
rs1031388567 Chr.20: 53967180-53967180 on Build GRCh38
rs753329363 Chr.20: 53967087-53967087 on Build GRCh38
rs1010489028 Chr.20: 53967103-53967103 on Build GRCh38
rs765788854 Chr.20: 53967115-53967115 on Build GRCh38
rs200990762 Chr.20: 53967119-53967119 on Build GRCh38
rs1034475938 Chr.20: 53967210-53967210 on Build GRCh38
rs370637556 Chr.20: 53967094-53967094 on Build GRCh38
rs1001311201 Chr.20: 53967198-53967198 on Build GRCh38
rs141548922 Chr.20: 53967163-53967163 on Build GRCh38
rs750971391 Chr.20: 53967116-53967116 on Build GRCh38
rs368202526 Chr.20: 53967067-53967067 on Build GRCh38
rs899725394 Chr.20: 53967096-53967096 on Build GRCh38
rs760299485 Chr.20: 53967081-53967081 on Build GRCh38
rs775115425 Chr.20: 53967077-53967077 on Build GRCh38
rs6097699 Chr.20: 53967149-53967149 on Build GRCh38
rs977252444 Chr.20: 53967178-53967178 on Build GRCh38
rs1029572297 Chr.20: 53967141-53967141 on Build GRCh38
rs758831601 Chr.20: 53967118-53967118 on Build GRCh38
rs572710510 Chr.20: 53967139-53967139 on Build GRCh38
rs763637376 Chr.20: 53967085-53967085 on Build GRCh38

View Full Product Details