Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 177436619-177436868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPS
Entrez Gene ID
8540
Gene Name
alkylglycerone phosphate synthase
Gene Aliases
ADAP-S, ADAS, ADHAPS, ADPS, ALDHPSY, RCDP3
UniGene
Hs.516543
Gene Chromosome Location
Chr.2: 177392743-177543836 on Build GRCh38
Transcripts
NM_003659.3 XM_011512041.2 XM_011512042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244651 c.545G>A p.R182Q Chr.2: 177436867-177436867 on Build GRCh38
1009932 c.447C>A p.S149S Chr.2: 177436769-177436769 on Build GRCh38
5699191 c.487G>T p.D163Y Chr.2: 177436809-177436809 on Build GRCh38
5343837 c.442-5T>A p.? Chr.2: 177436759-177436759 on Build GRCh38
6596416 c.502C>A p.L168I Chr.2: 177436824-177436824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202162408 Chr.2: 177436721-177436721 on Build GRCh38
rs1046007927 Chr.2: 177436705-177436705 on Build GRCh38
rs368566340 Chr.2: 177436743-177436743 on Build GRCh38
rs953796960 Chr.2: 177436673-177436673 on Build GRCh38
rs748129984 Chr.2: 177436786-177436786 on Build GRCh38
rs199934108 Chr.2: 177436722-177436722 on Build GRCh38
rs568037213 Chr.2: 177436677-177436677 on Build GRCh38
rs533896477 Chr.2: 177436698-177436698 on Build GRCh38
rs539922029 Chr.2: 177436787-177436787 on Build GRCh38
rs1022495522 Chr.2: 177436652-177436652 on Build GRCh38
rs913605632 Chr.2: 177436693-177436693 on Build GRCh38
rs989233747 Chr.2: 177436687-177436687 on Build GRCh38
rs909783707 Chr.2: 177436768-177436768 on Build GRCh38
rs778073526 Chr.2: 177436819-177436819 on Build GRCh38
rs886055164 Chr.2: 177436773-177436773 on Build GRCh38
rs3754989 Chr.2: 177436700-177436700 on Build GRCh38
rs201462821 Chr.2: 177436816-177436816 on Build GRCh38
rs371309373 Chr.2: 177436658-177436658 on Build GRCh38
rs755410191 Chr.2: 177436748-177436748 on Build GRCh38
rs972561409 Chr.2: 177436716-177436716 on Build GRCh38
rs780388817 Chr.2: 177436734-177436734 on Build GRCh38
rs116237975 Chr.2: 177436676-177436676 on Build GRCh38
rs781748049 Chr.2: 177436783-177436783 on Build GRCh38

View Full Product Details