Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 25680889-25681114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP10A
Entrez Gene ID
57194
Gene Name
ATPase phospholipid transporting 10A (putative)
Gene Aliases
ATP10C, ATPVA, ATPVC
UniGene
Hs.638517
Gene Chromosome Location
Chr.15: 25672241-25865144 on Build GRCh38
Transcripts
NM_024490.3 XM_005268261.4 XM_011521826.2 XM_011521828.2 XM_011521829.2 XM_011521830.2 XM_017022436.1 XM_017022437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
292367 c.3510G>C p.T1170T Chr.15: 25681057-25681057 on Build GRCh38
289829 c.3591C>T p.N1197N Chr.15: 25680897-25680897 on Build GRCh38
5054213 c.3510G>A p.T1170T Chr.15: 25681057-25681057 on Build GRCh38
3377523 c.3505C>T p.R1169* Chr.15: 25681062-25681062 on Build GRCh38
357236 c.3595G>T p.D1199Y Chr.15: 25680893-25680893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981172534 Chr.15: 25681043-25681043 on Build GRCh38
rs762306880 Chr.15: 25681038-25681038 on Build GRCh38
rs745420832 Chr.15: 25681083-25681083 on Build GRCh38
rs755033971 Chr.15: 25681058-25681058 on Build GRCh38
rs778814293 Chr.15: 25680951-25680951 on Build GRCh38
rs771825351 Chr.15: 25680997-25680997 on Build GRCh38
rs997221932 Chr.15: 25680995-25680995 on Build GRCh38
rs754910264 Chr.15: 25680937-25680937 on Build GRCh38
rs2076744 Chr.15: 25681032-25681032 on Build GRCh38
rs199878635 Chr.15: 25680929-25680929 on Build GRCh38
rs2076743 Chr.15: 25681039-25681039 on Build GRCh38
rs2076745 Chr.15: 25681005-25681005 on Build GRCh38
rs758287131 Chr.15: 25681067-25681067 on Build GRCh38
rs750750142 Chr.15: 25681050-25681050 on Build GRCh38
rs540435530 Chr.15: 25681011-25681011 on Build GRCh38
rs770556543 Chr.15: 25681071-25681071 on Build GRCh38
rs772955267 Chr.15: 25680998-25680998 on Build GRCh38
rs749617960 Chr.15: 25680951-25680951 on Build GRCh38
rs369411882 Chr.15: 25680950-25680950 on Build GRCh38
rs759084161 Chr.15: 25681033-25681033 on Build GRCh38
rs188478723 Chr.15: 25681068-25681068 on Build GRCh38
rs763910474 Chr.15: 25681007-25681007 on Build GRCh38
rs369583706 Chr.15: 25681082-25681082 on Build GRCh38
rs147170590 Chr.15: 25680932-25680932 on Build GRCh38
rs140438730 Chr.15: 25681061-25681061 on Build GRCh38
rs371481164 Chr.15: 25681085-25681085 on Build GRCh38
rs747992646 Chr.15: 25680983-25680983 on Build GRCh38
rs991527841 Chr.15: 25681062-25681062 on Build GRCh38
rs764757185 Chr.15: 25681035-25681035 on Build GRCh38
rs756318564 Chr.15: 25680917-25680917 on Build GRCh38
rs2076741 Chr.15: 25681057-25681057 on Build GRCh38
rs545631165 Chr.15: 25681036-25681036 on Build GRCh38
rs746751548 Chr.15: 25681070-25681070 on Build GRCh38
rs771205932 Chr.15: 25681084-25681084 on Build GRCh38
rs564882602 Chr.15: 25681022-25681022 on Build GRCh38
rs144251006 Chr.15: 25681018-25681018 on Build GRCh38
rs748117534 Chr.15: 25681065-25681065 on Build GRCh38
rs2076742 Chr.15: 25681051-25681051 on Build GRCh38

View Full Product Details