Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57802454-57802727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVIL
Entrez Gene ID
10677
Gene Name
advillin
Gene Aliases
ADVIL, DOC6, p92
UniGene
Hs.584854
Gene Chromosome Location
Chr.12: 57797372-57818704 on Build GRCh38
Transcripts
NM_006576.3 XM_017018710.1 XM_017018711.1 XM_017018712.1 XM_017018713.1 XM_017018714.1 XM_017018715.1 NM_001172697.1

Gene Symbol
TSFM
Entrez Gene ID
10102
Gene Name
Ts translation elongation factor, mitochondrial
Gene Aliases
EFTS, EFTSMT
UniGene
Hs.632704
Gene Chromosome Location
Chr.12: 57782745-57802856 on Build GRCh38
Transcripts
NM_006576.3 XM_017018710.1 XM_017018711.1 XM_017018712.1 XM_017018713.1 XM_017018714.1 XM_017018715.1 NM_001172697.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5774099 c.590G>A p.G197E Chr.12: 57802573-57802573 on Build GRCh38
1628753 c.608A>T p.Q203L Chr.12: 57802591-57802591 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs10783847 Chr.12: 57802664-57802664 on Build GRCh38
rs1016281716 Chr.12: 57802614-57802614 on Build GRCh38
rs1029779263 Chr.12: 57802616-57802616 on Build GRCh38
rs186621675 Chr.12: 57802574-57802574 on Build GRCh38
rs897306990 Chr.12: 57802639-57802639 on Build GRCh38
rs1024923597 Chr.12: 57802692-57802692 on Build GRCh38
rs1027003213 Chr.12: 57802660-57802660 on Build GRCh38
rs954265264 Chr.12: 57802629-57802629 on Build GRCh38
rs767822756 Chr.12: 57802511-57802511 on Build GRCh38
rs752861604 Chr.12: 57802541-57802541 on Build GRCh38
rs752174625 Chr.12: 57802478-57802478 on Build GRCh38
rs957293920 Chr.12: 57802684-57802685 on Build GRCh38
rs757747027 Chr.12: 57802578-57802578 on Build GRCh38
rs990746493 Chr.12: 57802690-57802690 on Build GRCh38
rs761089097 Chr.12: 57802559-57802559 on Build GRCh38
rs1051433094 Chr.12: 57802553-57802553 on Build GRCh38
rs1033843791 Chr.12: 57802505-57802505 on Build GRCh38
rs764456179 Chr.12: 57802642-57802642 on Build GRCh38
rs961375547 Chr.12: 57802585-57802585 on Build GRCh38
rs998573144 Chr.12: 57802606-57802606 on Build GRCh38

View Full Product Details