Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200608899-200609172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX1
Entrez Gene ID
316
Gene Name
aldehyde oxidase 1
Gene Aliases
AO, AOH1
UniGene
Hs.406238
Gene Chromosome Location
Chr.2: 200585952-200677064 on Build GRCh38
Transcripts
NM_001159.3 XM_011511062.1 XM_017003946.1 XM_017003947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
372223 c.948G>C p.K316N Chr.2: 200609024-200609024 on Build GRCh38
5914867 c.1049G>A p.R350K Chr.2: 200609125-200609125 on Build GRCh38
6090106 c.958G>T p.A320S Chr.2: 200609034-200609034 on Build GRCh38
5908063 c.928C>T p.L310F Chr.2: 200609004-200609004 on Build GRCh38
6239289 c.1027A>G p.T343A Chr.2: 200609103-200609103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35128788 Chr.2: 200609112-200609112 on Build GRCh38
rs756440136 Chr.2: 200609094-200609094 on Build GRCh38
rs371300618 Chr.2: 200609138-200609138 on Build GRCh38
rs780595492 Chr.2: 200609103-200609103 on Build GRCh38
rs777160401 Chr.2: 200609120-200609120 on Build GRCh38
rs780957516 Chr.2: 200609139-200609139 on Build GRCh38
rs886786724 Chr.2: 200608937-200608937 on Build GRCh38
rs750818785 Chr.2: 200609092-200609092 on Build GRCh38
rs113152957 Chr.2: 200609082-200609082 on Build GRCh38
rs762361811 Chr.2: 200609133-200609133 on Build GRCh38
rs908989769 Chr.2: 200609144-200609144 on Build GRCh38
rs764079026 Chr.2: 200609030-200609030 on Build GRCh38
rs755665118 Chr.2: 200609038-200609038 on Build GRCh38
rs58185012 Chr.2: 200609017-200609017 on Build GRCh38
rs969328982 Chr.2: 200608966-200608966 on Build GRCh38
rs746644279 Chr.2: 200608959-200608959 on Build GRCh38
rs765302652 Chr.2: 200608996-200608996 on Build GRCh38
rs751594946 Chr.2: 200609036-200609036 on Build GRCh38
rs757435420 Chr.2: 200609062-200609062 on Build GRCh38
rs759360483 Chr.2: 200608995-200608995 on Build GRCh38
rs571143352 Chr.2: 200609080-200609080 on Build GRCh38
rs776588487 Chr.2: 200608984-200608984 on Build GRCh38
rs780499300 Chr.2: 200608944-200608944 on Build GRCh38
rs775029057 Chr.2: 200609119-200609119 on Build GRCh38
rs765471606 Chr.2: 200609077-200609077 on Build GRCh38
rs372847747 Chr.2: 200608955-200608955 on Build GRCh38
rs975692005 Chr.2: 200609058-200609058 on Build GRCh38
rs770755882 Chr.2: 200609049-200609049 on Build GRCh38
rs960505211 Chr.2: 200609121-200609121 on Build GRCh38
rs2470902 Chr.2: 200608951-200608951 on Build GRCh38
rs148476608 Chr.2: 200609073-200609073 on Build GRCh38
rs374801878 Chr.2: 200608967-200608967 on Build GRCh38
rs534074509 Chr.2: 200608994-200608994 on Build GRCh38
rs367779647 Chr.2: 200609129-200609129 on Build GRCh38
rs747697694 Chr.2: 200608950-200608950 on Build GRCh38
rs866416276 Chr.2: 200609086-200609086 on Build GRCh38
rs772983456 Chr.2: 200608958-200608958 on Build GRCh38
rs763155474 Chr.2: 200609024-200609024 on Build GRCh38

View Full Product Details