Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38358183-38358444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERG
Entrez Gene ID
57828
Gene Name
cation channel sperm associated auxiliary subunit gamma
Gene Aliases
C19orf15
UniGene
Hs.324335
Gene Chromosome Location
Chr.19: 38335563-38370949 on Build GRCh38
Transcripts
NM_021185.4 XM_005259114.4 XM_005259115.4 XM_005259116.2 XM_006723308.3 XM_006723310.2 XM_006723311.2 XM_011527174.2 XM_011527175.2 XM_011527176.2 XM_011527177.2 XM_011527178.2 XM_011527179.1 XM_011527182.2 XM_017027053.1 XM_017027054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1726129 c.1295-2A>T p.? Chr.19: 38358276-38358276 on Build GRCh38
3737845 c.287-7C>T p.? Chr.19: 38358425-38358425 on Build GRCh38
3202078 c.271G>A p.E91K Chr.19: 38358313-38358313 on Build GRCh38
995929 c.250G>T p.E84* Chr.19: 38358292-38358292 on Build GRCh38
5916697 c.289C>T p.R97* Chr.19: 38358434-38358434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577229298 Chr.19: 38358426-38358426 on Build GRCh38
rs143271342 Chr.19: 38358312-38358312 on Build GRCh38
rs778862905 Chr.19: 38358369-38358369 on Build GRCh38
rs758117926 Chr.19: 38358378-38358378 on Build GRCh38
rs746512762 Chr.19: 38358376-38358376 on Build GRCh38
rs775816143 Chr.19: 38358425-38358425 on Build GRCh38
rs776155882 Chr.19: 38358357-38358357 on Build GRCh38
rs747735702 Chr.19: 38358377-38358377 on Build GRCh38
rs961798381 Chr.19: 38358263-38358263 on Build GRCh38
rs199974546 Chr.19: 38358315-38358315 on Build GRCh38
rs149594255 Chr.19: 38358285-38358285 on Build GRCh38
rs760298504 Chr.19: 38358302-38358302 on Build GRCh38
rs750459731 Chr.19: 38358347-38358347 on Build GRCh38
rs938389884 Chr.19: 38358297-38358297 on Build GRCh38
rs916181370 Chr.19: 38358308-38358308 on Build GRCh38
rs767849642 Chr.19: 38358341-38358341 on Build GRCh38
rs753857613 Chr.19: 38358354-38358354 on Build GRCh38
rs766901058 Chr.19: 38358307-38358307 on Build GRCh38
rs919147468 Chr.19: 38358407-38358407 on Build GRCh38
rs763269348 Chr.19: 38358313-38358313 on Build GRCh38
rs377193832 Chr.19: 38358389-38358389 on Build GRCh38
rs953306690 Chr.19: 38358205-38358205 on Build GRCh38
rs777538631 Chr.19: 38358380-38358380 on Build GRCh38
rs914975534 Chr.19: 38358330-38358330 on Build GRCh38
rs765598968 Chr.19: 38358316-38358317 on Build GRCh38
rs756267512 Chr.19: 38358348-38358348 on Build GRCh38
rs770186159 Chr.19: 38358303-38358303 on Build GRCh38
rs750312058 Chr.19: 38358251-38358251 on Build GRCh38
rs558788490 Chr.19: 38358337-38358337 on Build GRCh38
rs971841462 Chr.19: 38358271-38358271 on Build GRCh38
rs1012665052 Chr.19: 38358212-38358212 on Build GRCh38
rs369979949 Chr.19: 38358398-38358398 on Build GRCh38
rs201918503 Chr.19: 38358258-38358258 on Build GRCh38
rs763404052 Chr.19: 38358361-38358367 on Build GRCh38
rs372644714 Chr.19: 38358248-38358248 on Build GRCh38
rs780100264 Chr.19: 38358266-38358266 on Build GRCh38
rs754772093 Chr.19: 38358357-38358357 on Build GRCh38
rs749015283 Chr.19: 38358272-38358272 on Build GRCh38
rs771650795 Chr.19: 38358295-38358295 on Build GRCh38
rs772599567 Chr.19: 38358296-38358296 on Build GRCh38
rs768613205 Chr.19: 38358275-38358275 on Build GRCh38
rs751727297 Chr.19: 38358333-38358333 on Build GRCh38
rs8106895 Chr.19: 38358221-38358221 on Build GRCh38
rs770511935 Chr.19: 38358411-38358411 on Build GRCh38
rs372484319 Chr.19: 38358299-38358299 on Build GRCh38
rs971415502 Chr.19: 38358226-38358226 on Build GRCh38
rs764593398 Chr.19: 38358376-38358377 on Build GRCh38
rs779826154 Chr.19: 38358352-38358352 on Build GRCh38

View Full Product Details