Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2193560-2193813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAP20
Entrez Gene ID
199990
Gene Name
Fanconi anemia core complex associated protein 20
Gene Aliases
C1orf86, FP7162
UniGene
Hs.107101
Gene Chromosome Location
Chr.1: 2184460-2212720 on Build GRCh38
Transcripts
NM_001146310.1 NM_001256945.1 NM_001256946.1 NM_001256947.1 NM_001282670.1 NM_001282671.1 NM_001282672.1 NM_001282673.1 NM_182533.2 NR_046427.1 XM_006710419.3 XM_006710421.3 XM_011540914.2 XM_011540921.2 XM_011540922.1 XM_017000553.1 XM_017000554.1 XM_017000557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6019307 c.426G>A p.A142A Chr.1: 2193683-2193683 on Build GRCh38
5561953 c.716C>T p.P239L Chr.1: 2193702-2193702 on Build GRCh38
5705180 c.361C>T p.R121C Chr.1: 2193748-2193748 on Build GRCh38
6615331 c.462C>T p.F154F Chr.1: 2193647-2193647 on Build GRCh38
6855331 c.373G>A p.D125N Chr.1: 2193736-2193736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780196508 Chr.1: 2193767-2193767 on Build GRCh38
rs559286660 Chr.1: 2193586-2193586 on Build GRCh38
rs761368478 Chr.1: 2193609-2193609 on Build GRCh38
rs749586348 Chr.1: 2193650-2193650 on Build GRCh38
rs764940508 Chr.1: 2193744-2193744 on Build GRCh38
rs772593583 Chr.1: 2193794-2193794 on Build GRCh38
rs1047360016 Chr.1: 2193689-2193689 on Build GRCh38
rs748490576 Chr.1: 2193784-2193784 on Build GRCh38
rs377342456 Chr.1: 2193722-2193722 on Build GRCh38
rs760280006 Chr.1: 2193685-2193685 on Build GRCh38
rs779324738 Chr.1: 2193781-2193781 on Build GRCh38
rs746832136 Chr.1: 2193702-2193702 on Build GRCh38
rs942256490 Chr.1: 2193699-2193699 on Build GRCh38
rs765025402 Chr.1: 2193694-2193694 on Build GRCh38
rs113380716 Chr.1: 2193761-2193761 on Build GRCh38
rs773805992 Chr.1: 2193728-2193728 on Build GRCh38
rs774949618 Chr.1: 2193665-2193665 on Build GRCh38
rs753552259 Chr.1: 2193620-2193620 on Build GRCh38
rs200582608 Chr.1: 2193599-2193599 on Build GRCh38
rs762789045 Chr.1: 2193748-2193748 on Build GRCh38
rs748580410 Chr.1: 2193723-2193723 on Build GRCh38
rs755237245 Chr.1: 2193778-2193778 on Build GRCh38
rs1058411 Chr.1: 2193733-2193733 on Build GRCh38
rs754839843 Chr.1: 2193624-2193624 on Build GRCh38
rs981154294 Chr.1: 2193644-2193644 on Build GRCh38
rs765930009 Chr.1: 2193663-2193664 on Build GRCh38
rs144632131 Chr.1: 2193686-2193686 on Build GRCh38
rs752478314 Chr.1: 2193746-2193746 on Build GRCh38
rs200413325 Chr.1: 2193766-2193766 on Build GRCh38
rs781258049 Chr.1: 2193641-2193641 on Build GRCh38
rs772681827 Chr.1: 2193724-2193724 on Build GRCh38
rs772771641 Chr.1: 2193680-2193680 on Build GRCh38
rs957638292 Chr.1: 2193755-2193755 on Build GRCh38
rs756259328 Chr.1: 2193701-2193701 on Build GRCh38
rs373336716 Chr.1: 2193705-2193705 on Build GRCh38
rs753932361 Chr.1: 2193768-2193768 on Build GRCh38
rs998793719 Chr.1: 2193583-2193583 on Build GRCh38
rs751549543 Chr.1: 2193754-2193754 on Build GRCh38
rs777748430 Chr.1: 2193698-2193698 on Build GRCh38
rs201582904 Chr.1: 2193602-2193602 on Build GRCh38
rs138531306 Chr.1: 2193696-2193696 on Build GRCh38
rs750029490 Chr.1: 2193611-2193611 on Build GRCh38
rs377224564 Chr.1: 2193625-2193625 on Build GRCh38
rs746029487 Chr.1: 2193642-2193642 on Build GRCh38
rs775039498 Chr.1: 2193603-2193603 on Build GRCh38
rs750449732 Chr.1: 2193700-2193700 on Build GRCh38
rs113218107 Chr.1: 2193667-2193667 on Build GRCh38
rs998875403 Chr.1: 2193753-2193753 on Build GRCh38
rs757273561 Chr.1: 2193631-2193631 on Build GRCh38
rs762414864 Chr.1: 2193678-2193678 on Build GRCh38
rs769175099 Chr.1: 2193659-2193659 on Build GRCh38
rs375719713 Chr.1: 2193615-2193615 on Build GRCh38
rs763989535 Chr.1: 2193751-2193751 on Build GRCh38
rs759235931 Chr.1: 2193741-2193741 on Build GRCh38
rs114983157 Chr.1: 2193647-2193647 on Build GRCh38
rs759199804 Chr.1: 2193692-2193692 on Build GRCh38
rs753555895 Chr.1: 2193690-2193690 on Build GRCh38
rs769085065 Chr.1: 2193710-2193710 on Build GRCh38
rs776000071 Chr.1: 2193701-2193702 on Build GRCh38
rs776085784 Chr.1: 2193739-2193739 on Build GRCh38
rs760060398 Chr.1: 2193730-2193730 on Build GRCh38
rs200185087 Chr.1: 2193684-2193684 on Build GRCh38

View Full Product Details