Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 723718-723981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC78
Entrez Gene ID
124093
Gene Name
coiled-coil domain containing 78
Gene Aliases
C16orf25, CNM4, JFP10, hsCCDC78
UniGene
Hs.381943
Gene Chromosome Location
Chr.16: 722578-726880 on Build GRCh38
Transcripts
NM_001031737.2 XM_006720838.1 XM_006720843.3 XM_011522356.1 XM_011522357.1 XM_011522358.2 XM_011522359.1 XM_011522360.1 XM_011522361.1 XM_011522362.1 XM_011522363.1 XM_011522364.1 XM_011522365.1 XM_011522366.1 XM_011522367.1 XM_011522368.1 XM_011522369.1 XM_011522370.1 XM_011522371.2 XM_017022929.1 XM_017022930.1 XM_017022931.1 XM_017022932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
460447 c.1133+2T>C p.? Chr.16: 723855-723855 on Build GRCh38
6490671 c.1091delA p.K364fs*53 Chr.16: 723899-723899 on Build GRCh38
3691187 c.1080A>G p.S360S Chr.16: 723910-723910 on Build GRCh38
4521512 c.1115G>A p.G372E Chr.16: 723875-723875 on Build GRCh38
3387615 c.1057G>A p.G353S Chr.16: 723933-723933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763047897 Chr.16: 723955-723955 on Build GRCh38
rs756361553 Chr.16: 723809-723809 on Build GRCh38
rs929618078 Chr.16: 723843-723843 on Build GRCh38
rs199979958 Chr.16: 723951-723951 on Build GRCh38
rs776814118 Chr.16: 723893-723893 on Build GRCh38
rs759584555 Chr.16: 723840-723840 on Build GRCh38
rs906009986 Chr.16: 723956-723956 on Build GRCh38
rs145971446 Chr.16: 723887-723887 on Build GRCh38
rs766543879 Chr.16: 723827-723827 on Build GRCh38
rs370950546 Chr.16: 723903-723903 on Build GRCh38
rs780951516 Chr.16: 723867-723867 on Build GRCh38
rs769082660 Chr.16: 723959-723959 on Build GRCh38
rs557567837 Chr.16: 723884-723884 on Build GRCh38
rs921769306 Chr.16: 723908-723908 on Build GRCh38
rs907111705 Chr.16: 723885-723885 on Build GRCh38
rs752520956 Chr.16: 723819-723819 on Build GRCh38
rs1020256693 Chr.16: 723768-723770 on Build GRCh38
rs774132010 Chr.16: 723820-723820 on Build GRCh38
rs751900643 Chr.16: 723916-723916 on Build GRCh38
rs933251369 Chr.16: 723911-723911 on Build GRCh38
rs781457535 Chr.16: 723945-723945 on Build GRCh38
rs780209156 Chr.16: 723876-723876 on Build GRCh38
rs762323383 Chr.16: 723960-723960 on Build GRCh38
rs763378541 Chr.16: 723907-723907 on Build GRCh38
rs995716577 Chr.16: 723785-723785 on Build GRCh38
rs751889492 Chr.16: 723852-723852 on Build GRCh38
rs767325446 Chr.16: 723920-723920 on Build GRCh38
rs776726348 Chr.16: 723836-723836 on Build GRCh38
rs768334627 Chr.16: 723816-723816 on Build GRCh38
rs773310049 Chr.16: 723931-723931 on Build GRCh38
rs745320430 Chr.16: 723760-723760 on Build GRCh38
rs748870674 Chr.16: 723899-723900 on Build GRCh38
rs758861885 Chr.16: 723847-723847 on Build GRCh38
rs539475805 Chr.16: 723872-723872 on Build GRCh38
rs745409583 Chr.16: 723871-723871 on Build GRCh38
rs377343810 Chr.16: 723899-723899 on Build GRCh38
rs140104512 Chr.16: 723892-723892 on Build GRCh38
rs987374804 Chr.16: 723915-723915 on Build GRCh38
rs759668390 Chr.16: 723894-723894 on Build GRCh38
rs371958189 Chr.16: 723934-723934 on Build GRCh38
rs756867116 Chr.16: 723738-723738 on Build GRCh38
rs568871697 Chr.16: 723856-723856 on Build GRCh38
rs575159946 Chr.16: 723950-723950 on Build GRCh38
rs764612572 Chr.16: 723913-723913 on Build GRCh38
rs746094662 Chr.16: 723791-723791 on Build GRCh38
rs775450233 Chr.16: 723952-723952 on Build GRCh38
rs750098771 Chr.16: 723922-723922 on Build GRCh38
rs1021626011 Chr.16: 723891-723891 on Build GRCh38
rs1037367865 Chr.16: 723924-723924 on Build GRCh38
rs754906673 Chr.16: 723933-723933 on Build GRCh38
rs778396629 Chr.16: 723892-723892 on Build GRCh38
rs367596066 Chr.16: 723801-723801 on Build GRCh38
rs760821623 Chr.16: 723826-723826 on Build GRCh38
rs374450678 Chr.16: 723850-723850 on Build GRCh38
rs775570221 Chr.16: 723904-723904 on Build GRCh38
rs747939934 Chr.16: 723844-723844 on Build GRCh38
rs535293326 Chr.16: 723800-723800 on Build GRCh38
rs199867890 Chr.16: 723930-723930 on Build GRCh38
rs755798886 Chr.16: 723877-723877 on Build GRCh38
rs371542042 Chr.16: 723939-723939 on Build GRCh38
rs775534623 Chr.16: 723808-723808 on Build GRCh38
rs778741526 Chr.16: 723841-723841 on Build GRCh38
rs765483630 Chr.16: 723878-723878 on Build GRCh38
rs567996831 Chr.16: 723792-723792 on Build GRCh38
rs780859431 Chr.16: 723756-723756 on Build GRCh38
rs200482808 Chr.16: 723929-723929 on Build GRCh38
rs769879977 Chr.16: 723779-723779 on Build GRCh38

View Full Product Details