Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141279311-141279578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PXYLP1
Entrez Gene ID
92370
Gene Name
2-phosphoxylose phosphatase 1
Gene Aliases
ACPL2, HEL124, XYLP
UniGene
Hs.657887
Gene Chromosome Location
Chr.3: 141231825-141294903 on Build GRCh38
Transcripts
NM_001037172.2 NM_001282728.1 NM_152282.4 XM_005247903.3 XM_011513314.2 XM_017007510.1 XM_017007511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4319505 c.283C>T p.R95* Chr.3: 141279470-141279470 on Build GRCh38
4771983 c.243C>T p.H81H Chr.3: 141279430-141279430 on Build GRCh38
4771982 c.291C>T p.H97H Chr.3: 141279430-141279430 on Build GRCh38
3660225 c.245G>T p.G82V Chr.3: 141279432-141279432 on Build GRCh38
3720693 c.197C>G p.A66G Chr.3: 141279384-141279384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754829130 Chr.3: 141279426-141279426 on Build GRCh38
rs764812381 Chr.3: 141279542-141279543 on Build GRCh38
rs768347606 Chr.3: 141279473-141279473 on Build GRCh38
rs761163599 Chr.3: 141279510-141279510 on Build GRCh38
rs781330329 Chr.3: 141279445-141279445 on Build GRCh38
rs146572483 Chr.3: 141279471-141279471 on Build GRCh38
rs1028380792 Chr.3: 141279417-141279417 on Build GRCh38
rs1027457349 Chr.3: 141279494-141279494 on Build GRCh38
rs762789083 Chr.3: 141279524-141279524 on Build GRCh38
rs751853734 Chr.3: 141279542-141279542 on Build GRCh38
rs766136027 Chr.3: 141279529-141279529 on Build GRCh38
rs1035570616 Chr.3: 141279462-141279462 on Build GRCh38
rs761521004 Chr.3: 141279393-141279393 on Build GRCh38
rs774955602 Chr.3: 141279391-141279391 on Build GRCh38
rs374728196 Chr.3: 141279374-141279374 on Build GRCh38
rs748379587 Chr.3: 141279446-141279446 on Build GRCh38
rs773609750 Chr.3: 141279375-141279375 on Build GRCh38
rs1004566398 Chr.3: 141279551-141279551 on Build GRCh38
rs775047388 Chr.3: 141279459-141279459 on Build GRCh38
rs367938675 Chr.3: 141279387-141279387 on Build GRCh38
rs747123588 Chr.3: 141279361-141279361 on Build GRCh38
rs201688568 Chr.3: 141279392-141279392 on Build GRCh38
rs982291239 Chr.3: 141279457-141279457 on Build GRCh38
rs758150240 Chr.3: 141279403-141279403 on Build GRCh38
rs566040527 Chr.3: 141279521-141279521 on Build GRCh38
rs764740981 Chr.3: 141279394-141279394 on Build GRCh38
rs996856998 Chr.3: 141279398-141279398 on Build GRCh38
rs1040143705 Chr.3: 141279425-141279425 on Build GRCh38
rs766326945 Chr.3: 141279415-141279415 on Build GRCh38
rs756369533 Chr.3: 141279447-141279447 on Build GRCh38
rs893767926 Chr.3: 141279422-141279422 on Build GRCh38
rs13087169 Chr.3: 141279372-141279372 on Build GRCh38
rs750386405 Chr.3: 141279354-141279354 on Build GRCh38
rs764973527 Chr.3: 141279520-141279520 on Build GRCh38
rs776220175 Chr.3: 141279489-141279489 on Build GRCh38
rs199822582 Chr.3: 141279342-141279342 on Build GRCh38
rs200886677 Chr.3: 141279419-141279419 on Build GRCh38
rs199786824 Chr.3: 141279357-141279357 on Build GRCh38
rs78760647 Chr.3: 141279358-141279358 on Build GRCh38
rs757049989 Chr.3: 141279352-141279352 on Build GRCh38
rs777789397 Chr.3: 141279449-141279449 on Build GRCh38
rs763167344 Chr.3: 141279383-141279383 on Build GRCh38
rs778737327 Chr.3: 141279353-141279353 on Build GRCh38
rs750238675 Chr.3: 141279396-141279396 on Build GRCh38
rs200014913 Chr.3: 141279450-141279450 on Build GRCh38
rs901243870 Chr.3: 141279388-141279388 on Build GRCh38
rs755094963 Chr.3: 141279439-141279439 on Build GRCh38
rs751479859 Chr.3: 141279430-141279430 on Build GRCh38
rs768677037 Chr.3: 141279364-141279364 on Build GRCh38
rs375619423 Chr.3: 141279547-141279547 on Build GRCh38
rs771370657 Chr.3: 141279458-141279458 on Build GRCh38

View Full Product Details