Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79795033-79795258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX8
Entrez Gene ID
57332
Gene Name
chromobox 8
Gene Aliases
PC3, RC1
UniGene
Hs.387258
Gene Chromosome Location
Chr.17: 79794377-79797116 on Build GRCh38
Transcripts
NM_020649.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3523238 c.574C>T p.P192S Chr.17: 79795231-79795231 on Build GRCh38
6998890 c.696C>G p.D232E Chr.17: 79795109-79795109 on Build GRCh38
5004624 c.662G>A p.G221D Chr.17: 79795143-79795143 on Build GRCh38
6275568 c.582C>A p.D194E Chr.17: 79795223-79795223 on Build GRCh38
2802798 c.699C>T p.D233D Chr.17: 79795106-79795106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772859757 Chr.17: 79795186-79795186 on Build GRCh38
rs201171761 Chr.17: 79795145-79795145 on Build GRCh38
rs376248661 Chr.17: 79795109-79795109 on Build GRCh38
rs745874753 Chr.17: 79795086-79795086 on Build GRCh38
rs769294776 Chr.17: 79795235-79795235 on Build GRCh38
rs145608074 Chr.17: 79795096-79795096 on Build GRCh38
rs768020519 Chr.17: 79795139-79795139 on Build GRCh38
rs759875207 Chr.17: 79795137-79795137 on Build GRCh38
rs757296700 Chr.17: 79795205-79795205 on Build GRCh38
rs141960676 Chr.17: 79795104-79795104 on Build GRCh38
rs746994251 Chr.17: 79795226-79795226 on Build GRCh38
rs758300215 Chr.17: 79795218-79795218 on Build GRCh38
rs747621347 Chr.17: 79795232-79795232 on Build GRCh38
rs777797402 Chr.17: 79795144-79795144 on Build GRCh38
rs779967756 Chr.17: 79795222-79795222 on Build GRCh38
rs749429236 Chr.17: 79795058-79795058 on Build GRCh38
rs923018185 Chr.17: 79795093-79795093 on Build GRCh38
rs772768891 Chr.17: 79795238-79795238 on Build GRCh38
rs756214938 Chr.17: 79795143-79795143 on Build GRCh38
rs150395239 Chr.17: 79795148-79795148 on Build GRCh38
rs761146309 Chr.17: 79795198-79795198 on Build GRCh38
rs750611533 Chr.17: 79795217-79795217 on Build GRCh38
rs978886942 Chr.17: 79795212-79795212 on Build GRCh38
rs779016895 Chr.17: 79795209-79795209 on Build GRCh38
rs759913802 Chr.17: 79795191-79795191 on Build GRCh38
rs753857016 Chr.17: 79795200-79795200 on Build GRCh38
rs144276670 Chr.17: 79795156-79795156 on Build GRCh38
rs757406974 Chr.17: 79795146-79795146 on Build GRCh38
rs369528313 Chr.17: 79795195-79795195 on Build GRCh38
rs777990002 Chr.17: 79795054-79795054 on Build GRCh38
rs772133069 Chr.17: 79795088-79795088 on Build GRCh38
rs140970616 Chr.17: 79795163-79795163 on Build GRCh38
rs753325577 Chr.17: 79795136-79795137 on Build GRCh38
rs780825833 Chr.17: 79795230-79795230 on Build GRCh38
rs764216699 Chr.17: 79795199-79795199 on Build GRCh38
rs773995287 Chr.17: 79795077-79795077 on Build GRCh38
rs756127118 Chr.17: 79795052-79795052 on Build GRCh38
rs961666914 Chr.17: 79795228-79795228 on Build GRCh38
rs558160613 Chr.17: 79795181-79795181 on Build GRCh38
rs371617432 Chr.17: 79795106-79795106 on Build GRCh38
rs767825797 Chr.17: 79795194-79795194 on Build GRCh38
rs899259125 Chr.17: 79795142-79795142 on Build GRCh38
rs143582959 Chr.17: 79795102-79795102 on Build GRCh38
rs965280480 Chr.17: 79795169-79795169 on Build GRCh38
rs934938961 Chr.17: 79795070-79795070 on Build GRCh38
rs138089335 Chr.17: 79795097-79795097 on Build GRCh38
rs143202450 Chr.17: 79795182-79795182 on Build GRCh38
rs147538157 Chr.17: 79795185-79795185 on Build GRCh38
rs770798123 Chr.17: 79795076-79795076 on Build GRCh38
rs200482020 Chr.17: 79795229-79795229 on Build GRCh38
rs746904329 Chr.17: 79795157-79795157 on Build GRCh38
rs771935190 Chr.17: 79795152-79795152 on Build GRCh38
rs138228240 Chr.17: 79795151-79795151 on Build GRCh38

View Full Product Details