Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53715115-53715388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCOCO1
Entrez Gene ID
57658
Gene Name
calcium binding and coiled-coil domain 1
Gene Aliases
Cocoa, PP13275, calphoglin
UniGene
Hs.156667
Gene Chromosome Location
Chr.12: 53711118-53727523 on Build GRCh38
Transcripts
NM_001143682.1 NM_020898.2 NR_026554.1 XM_011538601.1 XM_011538602.1 XM_011538603.1 XM_011538604.1 XM_017019707.1 XM_017019708.1 XM_017019709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1705714 c.1303C>T p.R435* Chr.12: 53715283-53715283 on Build GRCh38
5516618 c.1187A>G p.K396R Chr.12: 53715213-53715213 on Build GRCh38
694064 c.1299A>G p.I433M Chr.12: 53715287-53715287 on Build GRCh38
4860570 c.1153A>G p.N385D Chr.12: 53715247-53715247 on Build GRCh38
5516617 c.1373A>G p.K458R Chr.12: 53715213-53715213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751725926 Chr.12: 53715212-53715212 on Build GRCh38
rs746605446 Chr.12: 53715282-53715282 on Build GRCh38
rs375019757 Chr.12: 53715338-53715338 on Build GRCh38
rs767455173 Chr.12: 53715334-53715334 on Build GRCh38
rs778714259 Chr.12: 53715270-53715270 on Build GRCh38
rs370012045 Chr.12: 53715158-53715158 on Build GRCh38
rs775884569 Chr.12: 53715298-53715298 on Build GRCh38
rs772727925 Chr.12: 53715161-53715161 on Build GRCh38
rs761952395 Chr.12: 53715332-53715332 on Build GRCh38
rs974961565 Chr.12: 53715353-53715353 on Build GRCh38
rs369001678 Chr.12: 53715235-53715235 on Build GRCh38
rs750540772 Chr.12: 53715221-53715221 on Build GRCh38
rs551055435 Chr.12: 53715219-53715219 on Build GRCh38
rs913876385 Chr.12: 53715179-53715179 on Build GRCh38
rs138639329 Chr.12: 53715311-53715311 on Build GRCh38
rs968891164 Chr.12: 53715352-53715352 on Build GRCh38
rs768752013 Chr.12: 53715265-53715265 on Build GRCh38
rs565082593 Chr.12: 53715209-53715209 on Build GRCh38
rs549518771 Chr.12: 53715177-53715177 on Build GRCh38
rs764564567 Chr.12: 53715210-53715210 on Build GRCh38
rs763297898 Chr.12: 53715308-53715308 on Build GRCh38
rs372812036 Chr.12: 53715220-53715220 on Build GRCh38
rs34311341 Chr.12: 53715216-53715216 on Build GRCh38
rs978545984 Chr.12: 53715336-53715336 on Build GRCh38
rs753754839 Chr.12: 53715356-53715356 on Build GRCh38
rs747752715 Chr.12: 53715277-53715277 on Build GRCh38
rs186570611 Chr.12: 53715335-53715335 on Build GRCh38
rs374769405 Chr.12: 53715229-53715229 on Build GRCh38
rs756452744 Chr.12: 53715337-53715337 on Build GRCh38
rs867989671 Chr.12: 53715195-53715195 on Build GRCh38
rs1002839709 Chr.12: 53715309-53715309 on Build GRCh38
rs974992848 Chr.12: 53715137-53715137 on Build GRCh38
rs376725569 Chr.12: 53715151-53715151 on Build GRCh38
rs898897091 Chr.12: 53715240-53715240 on Build GRCh38
rs757906844 Chr.12: 53715342-53715342 on Build GRCh38
rs933337990 Chr.12: 53715193-53715193 on Build GRCh38
rs748189250 Chr.12: 53715289-53715289 on Build GRCh38
rs201013720 Chr.12: 53715175-53715175 on Build GRCh38
rs754808822 Chr.12: 53715365-53715365 on Build GRCh38
rs1041781306 Chr.12: 53715163-53715163 on Build GRCh38
rs369142899 Chr.12: 53715362-53715362 on Build GRCh38
rs1045037986 Chr.12: 53715286-53715287 on Build GRCh38
rs144779779 Chr.12: 53715280-53715280 on Build GRCh38
rs1014343523 Chr.12: 53715328-53715328 on Build GRCh38
rs201669775 Chr.12: 53715200-53715200 on Build GRCh38
rs536673225 Chr.12: 53715295-53715295 on Build GRCh38
rs759959140 Chr.12: 53715174-53715174 on Build GRCh38
rs771426941 Chr.12: 53715279-53715279 on Build GRCh38
rs753238359 Chr.12: 53715185-53715185 on Build GRCh38
rs774658897 Chr.12: 53715317-53715317 on Build GRCh38
rs745344444 Chr.12: 53715184-53715184 on Build GRCh38
rs878862250 Chr.12: 53715167-53715167 on Build GRCh38
rs200012009 Chr.12: 53715288-53715288 on Build GRCh38

View Full Product Details