Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75640708-75640963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECQL5
Entrez Gene ID
9400
Gene Name
RecQ like helicase 5
Gene Aliases
RECQ5
UniGene
Hs.632229
Gene Chromosome Location
Chr.17: 75626845-75667202 on Build GRCh38
Transcripts
NM_004259.6 XM_005257818.3 XM_005257823.3 XM_011525482.2 XM_011525486.2 XM_017025343.1 NM_001162995.2 NR_028439.1 XM_011525118.2 XM_011525119.2 XM_017024945.1 XM_017024946.1 XM_017024947.1 XM_017024948.1

Gene Symbol
SMIM5
Entrez Gene ID
643008
Gene Name
small integral membrane protein 5
Gene Aliases
C17orf109, PP12104
UniGene
Hs.528605
Gene Chromosome Location
Chr.17: 75633434-75641406 on Build GRCh38
Transcripts
NM_004259.6 XM_005257818.3 XM_005257823.3 XM_011525482.2 XM_011525486.2 XM_017025343.1 NM_001162995.2 NR_028439.1 XM_011525118.2 XM_011525119.2 XM_017024945.1 XM_017024946.1 XM_017024947.1 XM_017024948.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6319044 c.167G>T p.C56F Chr.17: 75640830-75640830 on Build GRCh38
7289905 c.134T>C p.V45A Chr.17: 75640797-75640797 on Build GRCh38
7312661 c.150G>A p.L50L Chr.17: 75640813-75640813 on Build GRCh38
400538 c.224C>A p.T75K Chr.17: 75640887-75640887 on Build GRCh38
7329983 c.141G>T p.L47L Chr.17: 75640804-75640804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910383988 Chr.17: 75640847-75640847 on Build GRCh38
rs772747227 Chr.17: 75640911-75640911 on Build GRCh38
rs375715008 Chr.17: 75640780-75640780 on Build GRCh38
rs929029254 Chr.17: 75640891-75640891 on Build GRCh38
rs779858935 Chr.17: 75640788-75640788 on Build GRCh38
rs1047435169 Chr.17: 75640914-75640916 on Build GRCh38
rs756859009 Chr.17: 75640761-75640761 on Build GRCh38
rs201220906 Chr.17: 75640879-75640879 on Build GRCh38
rs72854944 Chr.17: 75640781-75640781 on Build GRCh38
rs1012899518 Chr.17: 75640791-75640791 on Build GRCh38
rs761285592 Chr.17: 75640903-75640903 on Build GRCh38
rs888727816 Chr.17: 75640735-75640735 on Build GRCh38
rs573377672 Chr.17: 75640765-75640766 on Build GRCh38
rs565540953 Chr.17: 75640742-75640742 on Build GRCh38
rs1005784784 Chr.17: 75640737-75640737 on Build GRCh38
rs374319429 Chr.17: 75640792-75640792 on Build GRCh38
rs527721770 Chr.17: 75640743-75640743 on Build GRCh38
rs986035859 Chr.17: 75640838-75640838 on Build GRCh38
rs749256290 Chr.17: 75640865-75640865 on Build GRCh38
rs919041248 Chr.17: 75640885-75640885 on Build GRCh38
rs1031765759 Chr.17: 75640835-75640835 on Build GRCh38
rs1056017728 Chr.17: 75640904-75640904 on Build GRCh38
rs561273921 Chr.17: 75640768-75640768 on Build GRCh38
rs777283585 Chr.17: 75640769-75640769 on Build GRCh38
rs770328446 Chr.17: 75640826-75640827 on Build GRCh38
rs894650777 Chr.17: 75640907-75640907 on Build GRCh38
rs1034951551 Chr.17: 75640740-75640740 on Build GRCh38
rs990425609 Chr.17: 75640857-75640857 on Build GRCh38
rs370000834 Chr.17: 75640856-75640856 on Build GRCh38
rs746853896 Chr.17: 75640811-75640811 on Build GRCh38
rs887236824 Chr.17: 75640758-75640758 on Build GRCh38
rs775976120 Chr.17: 75640938-75640938 on Build GRCh38
rs925957719 Chr.17: 75640793-75640793 on Build GRCh38
rs778653203 Chr.17: 75640773-75640773 on Build GRCh38
rs991006267 Chr.17: 75640926-75640926 on Build GRCh38
rs1017178367 Chr.17: 75640767-75640767 on Build GRCh38
rs531373402 Chr.17: 75640902-75640902 on Build GRCh38
rs975761289 Chr.17: 75640895-75640895 on Build GRCh38
rs769987205 Chr.17: 75640853-75640853 on Build GRCh38
rs1040226941 Chr.17: 75640884-75640884 on Build GRCh38
rs77898201 Chr.17: 75640898-75640898 on Build GRCh38
rs957160871 Chr.17: 75640852-75640852 on Build GRCh38
rs116346711 Chr.17: 75640759-75640759 on Build GRCh38

View Full Product Details