Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 181651261-181651524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1E
Entrez Gene ID
777
Gene Name
calcium voltage-gated channel subunit alpha1 E
Gene Aliases
BII, CACH6, CACNL1A6, Cav2.3, gm139
UniGene
Hs.437444
Gene Chromosome Location
Chr.1: 181317712-181808084 on Build GRCh38
Transcripts
NM_000721.3 NM_001205293.1 NM_001205294.1 XM_017002243.1 XM_017002244.1 XM_017002245.1 XM_017002246.1 XM_017002247.1 XM_017002248.1 XM_017002249.1 XM_017002250.1 XM_017002251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4025749 c.988T>C p.Y330H Chr.1: 181651374-181651374 on Build GRCh38
5031406 c.1041G>A p.L347L Chr.1: 181651427-181651427 on Build GRCh38
6059819 c.838C>A p.L280M Chr.1: 181651371-181651371 on Build GRCh38
6249300 c.823G>T p.A275S Chr.1: 181651356-181651356 on Build GRCh38
4025752 c.991T>G p.F331V Chr.1: 181651377-181651377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006102872 Chr.1: 181651471-181651471 on Build GRCh38
rs544058072 Chr.1: 181651469-181651469 on Build GRCh38
rs745797769 Chr.1: 181651419-181651419 on Build GRCh38
rs200603172 Chr.1: 181651409-181651409 on Build GRCh38
rs913919274 Chr.1: 181651282-181651282 on Build GRCh38
rs746946402 Chr.1: 181651291-181651291 on Build GRCh38
rs764126261 Chr.1: 181651330-181651330 on Build GRCh38
rs766873635 Chr.1: 181651470-181651470 on Build GRCh38
rs201080304 Chr.1: 181651439-181651439 on Build GRCh38
rs756614242 Chr.1: 181651371-181651371 on Build GRCh38
rs886039323 Chr.1: 181651440-181651440 on Build GRCh38
rs761924788 Chr.1: 181651449-181651449 on Build GRCh38
rs769612243 Chr.1: 181651421-181651421 on Build GRCh38
rs555540452 Chr.1: 181651355-181651355 on Build GRCh38
rs746463249 Chr.1: 181651394-181651394 on Build GRCh38
rs897477762 Chr.1: 181651486-181651486 on Build GRCh38
rs147221625 Chr.1: 181651340-181651340 on Build GRCh38
rs754054442 Chr.1: 181651382-181651382 on Build GRCh38
rs775638047 Chr.1: 181651321-181651321 on Build GRCh38
rs1014959978 Chr.1: 181651488-181651488 on Build GRCh38
rs370260416 Chr.1: 181651450-181651450 on Build GRCh38
rs754194469 Chr.1: 181651472-181651472 on Build GRCh38
rs777316858 Chr.1: 181651289-181651289 on Build GRCh38
rs769560869 Chr.1: 181651319-181651319 on Build GRCh38
rs118043317 Chr.1: 181651453-181651453 on Build GRCh38
rs779874299 Chr.1: 181651424-181651424 on Build GRCh38
rs776420019 Chr.1: 181651299-181651299 on Build GRCh38
rs758021122 Chr.1: 181651385-181651385 on Build GRCh38
rs745607997 Chr.1: 181651307-181651307 on Build GRCh38
rs780852075 Chr.1: 181651342-181651342 on Build GRCh38
rs757203951 Chr.1: 181651403-181651403 on Build GRCh38
rs761243813 Chr.1: 181651460-181651460 on Build GRCh38
rs768069842 Chr.1: 181651361-181651361 on Build GRCh38
rs763200598 Chr.1: 181651325-181651325 on Build GRCh38
rs762519058 Chr.1: 181651494-181651494 on Build GRCh38
rs777182809 Chr.1: 181651388-181651388 on Build GRCh38
rs372780939 Chr.1: 181651479-181651479 on Build GRCh38
rs770817679 Chr.1: 181651296-181651296 on Build GRCh38
rs574139937 Chr.1: 181651367-181651367 on Build GRCh38
rs753352249 Chr.1: 181651324-181651327 on Build GRCh38

View Full Product Details