Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26198004-26198243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER4
Entrez Gene ID
378807
Gene Name
cation channel sperm associated 4
Gene Aliases
-
UniGene
Hs.123532
Gene Chromosome Location
Chr.1: 26189962-26202542 on Build GRCh38
Transcripts
NM_198137.1 XM_011541432.2 XM_011541433.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3400638 c.670T>G p.F224V Chr.1: 26198069-26198069 on Build GRCh38
464350 c.678G>A p.L226L Chr.1: 26198077-26198077 on Build GRCh38
4030940 c.664C>A p.L222I Chr.1: 26198063-26198063 on Build GRCh38
5004895 c.615C>T p.R205R Chr.1: 26198014-26198014 on Build GRCh38
4751528 c.634C>T p.P212S Chr.1: 26198033-26198033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749125170 Chr.1: 26198097-26198097 on Build GRCh38
rs776229141 Chr.1: 26198128-26198128 on Build GRCh38
rs754218493 Chr.1: 26198051-26198051 on Build GRCh38
rs200696714 Chr.1: 26198121-26198121 on Build GRCh38
rs202016961 Chr.1: 26198094-26198094 on Build GRCh38
rs960458602 Chr.1: 26198189-26198189 on Build GRCh38
rs35577175 Chr.1: 26198188-26198189 on Build GRCh38
rs750934408 Chr.1: 26198071-26198071 on Build GRCh38
rs368402192 Chr.1: 26198093-26198093 on Build GRCh38
rs199965982 Chr.1: 26198125-26198125 on Build GRCh38
rs61776651 Chr.1: 26198029-26198029 on Build GRCh38
rs9970046 Chr.1: 26198215-26198215 on Build GRCh38
rs770890331 Chr.1: 26198098-26198098 on Build GRCh38
rs761887602 Chr.1: 26198128-26198128 on Build GRCh38
rs17163655 Chr.1: 26198065-26198065 on Build GRCh38
rs562179031 Chr.1: 26198207-26198207 on Build GRCh38
rs760122372 Chr.1: 26198123-26198123 on Build GRCh38
rs776731511 Chr.1: 26198122-26198129 on Build GRCh38
rs757896068 Chr.1: 26198197-26198197 on Build GRCh38
rs879053345 Chr.1: 26198090-26198090 on Build GRCh38
rs772038788 Chr.1: 26198120-26198120 on Build GRCh38
rs745847390 Chr.1: 26198112-26198112 on Build GRCh38
rs542376854 Chr.1: 26198173-26198173 on Build GRCh38
rs560352853 Chr.1: 26198060-26198060 on Build GRCh38
rs138417334 Chr.1: 26198043-26198043 on Build GRCh38
rs780056886 Chr.1: 26198088-26198088 on Build GRCh38
rs775048760 Chr.1: 26198022-26198022 on Build GRCh38
rs577071850 Chr.1: 26198073-26198073 on Build GRCh38
rs747109687 Chr.1: 26198092-26198092 on Build GRCh38
rs774168824 Chr.1: 26198099-26198099 on Build GRCh38
rs764051573 Chr.1: 26198095-26198095 on Build GRCh38

View Full Product Details