Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 222693776-222694009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIDA
Entrez Gene ID
64853
Gene Name
axin interactor, dorsalization associated
Gene Aliases
C1orf80
UniGene
Hs.156625
Gene Chromosome Location
Chr.1: 222668013-222712522 on Build GRCh38
Transcripts
NM_022831.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4390360 c.173C>G p.S58C Chr.1: 222693833-222693833 on Build GRCh38
4390359 c.245C>G p.S82C Chr.1: 222693833-222693833 on Build GRCh38
463982 c.286C>T p.P96S Chr.1: 222693792-222693792 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781718432 Chr.1: 222693837-222693837 on Build GRCh38
rs774261135 Chr.1: 222693962-222693962 on Build GRCh38
rs969075534 Chr.1: 222693828-222693828 on Build GRCh38
rs761306133 Chr.1: 222693930-222693930 on Build GRCh38
rs1037441101 Chr.1: 222693872-222693885 on Build GRCh38
rs780431970 Chr.1: 222693851-222693851 on Build GRCh38
rs750947055 Chr.1: 222693839-222693839 on Build GRCh38
rs945791002 Chr.1: 222693941-222693941 on Build GRCh38
rs768924874 Chr.1: 222693823-222693825 on Build GRCh38
rs771106788 Chr.1: 222693891-222693891 on Build GRCh38
rs201118792 Chr.1: 222693860-222693860 on Build GRCh38
rs1041516716 Chr.1: 222693965-222693965 on Build GRCh38
rs749466629 Chr.1: 222693874-222693874 on Build GRCh38
rs992463116 Chr.1: 222693812-222693812 on Build GRCh38
rs775398868 Chr.1: 222693923-222693923 on Build GRCh38
rs1002098890 Chr.1: 222693889-222693889 on Build GRCh38
rs202083483 Chr.1: 222693869-222693869 on Build GRCh38
rs757636166 Chr.1: 222693833-222693833 on Build GRCh38
rs112965719 Chr.1: 222693950-222693950 on Build GRCh38
rs756473755 Chr.1: 222693841-222693841 on Build GRCh38
rs946216015 Chr.1: 222693978-222693978 on Build GRCh38
rs902663884 Chr.1: 222693967-222693967 on Build GRCh38
rs542389245 Chr.1: 222693831-222693831 on Build GRCh38
rs186791914 Chr.1: 222693894-222693894 on Build GRCh38
rs764789665 Chr.1: 222693824-222693824 on Build GRCh38
rs755209531 Chr.1: 222693877-222693877 on Build GRCh38
rs753508631 Chr.1: 222693823-222693823 on Build GRCh38
rs913366965 Chr.1: 222693906-222693906 on Build GRCh38
rs936369532 Chr.1: 222693896-222693896 on Build GRCh38
rs561681172 Chr.1: 222693904-222693904 on Build GRCh38

View Full Product Details