Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68250651-68250912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMZ2
Entrez Gene ID
51321
Gene Name
archaelysin family metallopeptidase 2
Gene Aliases
-
UniGene
Hs.293560
Gene Chromosome Location
Chr.17: 68205486-68257164 on Build GRCh38
Transcripts
NM_001033569.1 NM_001033570.1 NM_001033571.1 NM_001033572.1 NM_001033574.1 NM_001289054.1 NM_001289056.1 NM_016627.4 XM_005257432.4 XM_005257433.3 XM_005257435.4 XM_005257436.3 XM_006721940.2 XM_017024723.1 XM_017024724.1 XM_017024725.1 XM_017024726.1 XM_017024727.1 XM_017024728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4774214 c.385C>A p.P129T Chr.17: 68250895-68250895 on Build GRCh38
4068960 c.340T>G p.Y114D Chr.17: 68250850-68250850 on Build GRCh38
4774810 c.366G>T p.L122F Chr.17: 68250876-68250876 on Build GRCh38
7076606 c.358T>C p.Y120H Chr.17: 68250868-68250868 on Build GRCh38
3795991 c.357C>G p.F119L Chr.17: 68250867-68250867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781920066 Chr.17: 68250833-68250833 on Build GRCh38
rs782517683 Chr.17: 68250879-68250879 on Build GRCh38
rs782363962 Chr.17: 68250801-68250801 on Build GRCh38
rs200392154 Chr.17: 68250876-68250876 on Build GRCh38
rs782442645 Chr.17: 68250857-68250857 on Build GRCh38
rs782586964 Chr.17: 68250763-68250763 on Build GRCh38
rs191065828 Chr.17: 68250738-68250738 on Build GRCh38
rs782264957 Chr.17: 68250793-68250793 on Build GRCh38
rs200882036 Chr.17: 68250766-68250766 on Build GRCh38
rs781836023 Chr.17: 68250743-68250743 on Build GRCh38
rs782646946 Chr.17: 68250754-68250754 on Build GRCh38
rs201005237 Chr.17: 68250845-68250845 on Build GRCh38
rs782649154 Chr.17: 68250886-68250886 on Build GRCh38
rs782752418 Chr.17: 68250877-68250877 on Build GRCh38
rs782208686 Chr.17: 68250787-68250787 on Build GRCh38
rs548277332 Chr.17: 68250699-68250699 on Build GRCh38
rs782370345 Chr.17: 68250785-68250785 on Build GRCh38
rs782598215 Chr.17: 68250789-68250789 on Build GRCh38
rs782539113 Chr.17: 68250757-68250757 on Build GRCh38
rs563962128 Chr.17: 68250720-68250720 on Build GRCh38
rs200407287 Chr.17: 68250834-68250834 on Build GRCh38
rs560380503 Chr.17: 68250713-68250714 on Build GRCh38
rs781909565 Chr.17: 68250756-68250756 on Build GRCh38
rs782461327 Chr.17: 68250751-68250751 on Build GRCh38
rs61748823 Chr.17: 68250820-68250820 on Build GRCh38
rs781811392 Chr.17: 68250870-68250870 on Build GRCh38
rs369248902 Chr.17: 68250716-68250716 on Build GRCh38
rs781830885 Chr.17: 68250878-68250878 on Build GRCh38
rs782740642 Chr.17: 68250836-68250836 on Build GRCh38
rs183324449 Chr.17: 68250846-68250846 on Build GRCh38
rs782319149 Chr.17: 68250831-68250831 on Build GRCh38
rs782790644 Chr.17: 68250853-68250853 on Build GRCh38
rs782029483 Chr.17: 68250819-68250819 on Build GRCh38

View Full Product Details