Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81190526-81190759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP131
Entrez Gene ID
22994
Gene Name
centrosomal protein 131
Gene Aliases
AZ1, AZI1, ZA1
UniGene
Hs.514578
Gene Chromosome Location
Chr.17: 81189593-81222989 on Build GRCh38
Transcripts
NM_001009811.3 NM_001319228.1 NM_001319229.1 NM_014984.3 XM_011524547.2 XM_011524548.2 XM_011524550.2 XM_011524551.2 XM_011524552.2 XM_011524553.2 XM_011524555.2 XM_017024369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086530 c.2917G>A p.E973K Chr.17: 81190721-81190721 on Build GRCh38
50447 c.2954C>T p.T985M Chr.17: 81190684-81190684 on Build GRCh38
708675 c.2892G>T p.E964D Chr.17: 81190746-81190746 on Build GRCh38
6225714 c.2884C>T p.R962C Chr.17: 81190754-81190754 on Build GRCh38
5762766 c.2991C>T p.I997I Chr.17: 81190755-81190755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528361973 Chr.17: 81190609-81190609 on Build GRCh38
rs776115559 Chr.17: 81190695-81190695 on Build GRCh38
rs777884107 Chr.17: 81190676-81190676 on Build GRCh38
rs761040753 Chr.17: 81190604-81190604 on Build GRCh38
rs749204255 Chr.17: 81190628-81190628 on Build GRCh38
rs766797351 Chr.17: 81190647-81190647 on Build GRCh38
rs763456299 Chr.17: 81190697-81190697 on Build GRCh38
rs112626893 Chr.17: 81190590-81190590 on Build GRCh38
rs775722037 Chr.17: 81190599-81190600 on Build GRCh38
rs751942226 Chr.17: 81190708-81190708 on Build GRCh38
rs767023822 Chr.17: 81190632-81190632 on Build GRCh38
rs947863115 Chr.17: 81190668-81190668 on Build GRCh38
rs747290866 Chr.17: 81190604-81190604 on Build GRCh38
rs190069768 Chr.17: 81190608-81190608 on Build GRCh38
rs745613456 Chr.17: 81190684-81190684 on Build GRCh38
rs766813702 Chr.17: 81190698-81190698 on Build GRCh38
rs755256899 Chr.17: 81190709-81190709 on Build GRCh38
rs1026558901 Chr.17: 81190592-81190592 on Build GRCh38
rs759325364 Chr.17: 81190630-81190630 on Build GRCh38
rs778063233 Chr.17: 81190725-81190725 on Build GRCh38
rs770984487 Chr.17: 81190683-81190683 on Build GRCh38
rs767890571 Chr.17: 81190711-81190711 on Build GRCh38
rs781222283 Chr.17: 81190671-81190671 on Build GRCh38
rs550696518 Chr.17: 81190717-81190717 on Build GRCh38
rs200888368 Chr.17: 81190642-81190642 on Build GRCh38
rs965503535 Chr.17: 81190589-81190589 on Build GRCh38
rs1019594667 Chr.17: 81190696-81190696 on Build GRCh38
rs756185994 Chr.17: 81190673-81190673 on Build GRCh38
rs772981057 Chr.17: 81190625-81190625 on Build GRCh38
rs117616373 Chr.17: 81190688-81190688 on Build GRCh38
rs749310378 Chr.17: 81190735-81190735 on Build GRCh38
rs568563543 Chr.17: 81190722-81190722 on Build GRCh38
rs895009375 Chr.17: 81190667-81190667 on Build GRCh38
rs748288587 Chr.17: 81190672-81190672 on Build GRCh38
rs770896458 Chr.17: 81190629-81190629 on Build GRCh38
rs371442789 Chr.17: 81190633-81190633 on Build GRCh38
rs915306585 Chr.17: 81190618-81190618 on Build GRCh38
rs779242406 Chr.17: 81190598-81190598 on Build GRCh38
rs946731436 Chr.17: 81190620-81190620 on Build GRCh38
rs750833778 Chr.17: 81190600-81190600 on Build GRCh38
rs755205585 Chr.17: 81190662-81190662 on Build GRCh38
rs778958570 Chr.17: 81190742-81190742 on Build GRCh38
rs201747795 Chr.17: 81190614-81190614 on Build GRCh38
rs769790630 Chr.17: 81190621-81190621 on Build GRCh38
rs760355681 Chr.17: 81190692-81190692 on Build GRCh38
rs757800182 Chr.17: 81190595-81190595 on Build GRCh38
rs763572417 Chr.17: 81190694-81190694 on Build GRCh38
rs1028551068 Chr.17: 81190597-81190597 on Build GRCh38
rs757286425 Chr.17: 81190736-81190736 on Build GRCh38
rs778272513 Chr.17: 81190603-81190603 on Build GRCh38
rs779477803 Chr.17: 81190653-81190653 on Build GRCh38
rs898242637 Chr.17: 81190674-81190674 on Build GRCh38
rs757751041 Chr.17: 81190643-81190643 on Build GRCh38
rs749292248 Chr.17: 81190679-81190679 on Build GRCh38
rs758804062 Chr.17: 81190602-81190602 on Build GRCh38

View Full Product Details