Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86385978-86386215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC81
Entrez Gene ID
60494
Gene Name
coiled-coil domain containing 81
Gene Aliases
-
UniGene
Hs.144913
Gene Chromosome Location
Chr.11: 86374736-86423109 on Build GRCh38
Transcripts
NM_001156474.1 NM_021827.4 XM_005274161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
467514 c.115G>A p.V39M Chr.11: 86386086-86386086 on Build GRCh38
467516 c.124C>T p.Q42* Chr.11: 86386095-86386095 on Build GRCh38
467515 c.123G>T p.R41R Chr.11: 86386094-86386094 on Build GRCh38
4641579 c.116T>C p.V39A Chr.11: 86386087-86386087 on Build GRCh38
3454105 c.131C>T p.T44I Chr.11: 86386102-86386102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527394882 Chr.11: 86386141-86386141 on Build GRCh38
rs371436469 Chr.11: 86386107-86386107 on Build GRCh38
rs370721972 Chr.11: 86386009-86386009 on Build GRCh38
rs1008560885 Chr.11: 86386119-86386119 on Build GRCh38
rs890160168 Chr.11: 86386094-86386094 on Build GRCh38
rs768790448 Chr.11: 86386069-86386069 on Build GRCh38
rs573776375 Chr.11: 86386118-86386125 on Build GRCh38
rs778403625 Chr.11: 86386112-86386112 on Build GRCh38
rs767691902 Chr.11: 86386074-86386074 on Build GRCh38
rs749584353 Chr.11: 86386148-86386148 on Build GRCh38
rs780352481 Chr.11: 86386144-86386144 on Build GRCh38
rs754600068 Chr.11: 86386019-86386019 on Build GRCh38
rs1018299940 Chr.11: 86386178-86386178 on Build GRCh38
rs769051037 Chr.11: 86386146-86386153 on Build GRCh38
rs757836534 Chr.11: 86386127-86386127 on Build GRCh38
rs775841655 Chr.11: 86386062-86386062 on Build GRCh38
rs564929766 Chr.11: 86386111-86386111 on Build GRCh38
rs572214522 Chr.11: 86386122-86386145 on Build GRCh38
rs747407773 Chr.11: 86386130-86386133 on Build GRCh38
rs958490035 Chr.11: 86386184-86386184 on Build GRCh38
rs760694239 Chr.11: 86386093-86386093 on Build GRCh38
rs761770076 Chr.11: 86386006-86386006 on Build GRCh38
rs368543445 Chr.11: 86386109-86386109 on Build GRCh38
rs1006950024 Chr.11: 86386164-86386164 on Build GRCh38
rs777266364 Chr.11: 86386136-86386136 on Build GRCh38
rs778438967 Chr.11: 86386024-86386024 on Build GRCh38
rs199553106 Chr.11: 86386010-86386010 on Build GRCh38
rs774674334 Chr.11: 86386072-86386072 on Build GRCh38
rs769112952 Chr.11: 86386152-86386152 on Build GRCh38
rs994965182 Chr.11: 86386147-86386147 on Build GRCh38
rs746435140 Chr.11: 86386056-86386056 on Build GRCh38
rs763182214 Chr.11: 86386067-86386067 on Build GRCh38
rs746314412 Chr.11: 86386138-86386138 on Build GRCh38
rs897559858 Chr.11: 86386145-86386145 on Build GRCh38
rs576072834 Chr.11: 86386086-86386086 on Build GRCh38
rs545760960 Chr.11: 86386045-86386045 on Build GRCh38
rs1027461732 Chr.11: 86386175-86386175 on Build GRCh38
rs771656666 Chr.11: 86386040-86386040 on Build GRCh38
rs752445220 Chr.11: 86386118-86386118 on Build GRCh38
rs1032170297 Chr.11: 86386149-86386149 on Build GRCh38
rs192546932 Chr.11: 86386013-86386013 on Build GRCh38
rs893956907 Chr.11: 86386030-86386030 on Build GRCh38
rs777102676 Chr.11: 86386150-86386153 on Build GRCh38
rs545045634 Chr.11: 86386092-86386092 on Build GRCh38
rs780421110 Chr.11: 86386121-86386122 on Build GRCh38
rs141858763 Chr.11: 86386073-86386073 on Build GRCh38
rs747647533 Chr.11: 86386025-86386025 on Build GRCh38
rs778914711 Chr.11: 86386054-86386055 on Build GRCh38
rs753475871 Chr.11: 86386014-86386014 on Build GRCh38
rs770075290 Chr.11: 86386059-86386059 on Build GRCh38
rs766219476 Chr.11: 86386103-86386103 on Build GRCh38

View Full Product Details