Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112116475-112116700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf52
Entrez Gene ID
79669
Gene Name
chromosome 3 open reading frame 52
Gene Aliases
TTMP
UniGene
Hs.434247
Gene Chromosome Location
Chr.3: 112086335-112131095 on Build GRCh38
Transcripts
NM_001171747.1 NM_024616.2 XM_011513133.2 XM_017007181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4845657 c.397-1G>C p.? Chr.3: 112116641-112116641 on Build GRCh38
5695613 c.401A>T p.E134V Chr.3: 112116646-112116646 on Build GRCh38
4845658 c.650-1G>C p.? Chr.3: 112116641-112116641 on Build GRCh38
5695614 c.654A>T p.*218C Chr.3: 112116646-112116646 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779014713 Chr.3: 112116619-112116619 on Build GRCh38
rs760580736 Chr.3: 112116661-112116661 on Build GRCh38
rs758669369 Chr.3: 112116657-112116657 on Build GRCh38
rs758613704 Chr.3: 112116651-112116651 on Build GRCh38
rs993258121 Chr.3: 112116519-112116519 on Build GRCh38
rs1026506300 Chr.3: 112116532-112116532 on Build GRCh38
rs781676502 Chr.3: 112116663-112116663 on Build GRCh38
rs1025089277 Chr.3: 112116595-112116595 on Build GRCh38
rs936783499 Chr.3: 112116672-112116672 on Build GRCh38
rs535352806 Chr.3: 112116560-112116560 on Build GRCh38
rs992375623 Chr.3: 112116580-112116580 on Build GRCh38
rs377500470 Chr.3: 112116534-112116534 on Build GRCh38
rs555976522 Chr.3: 112116618-112116618 on Build GRCh38
rs750171500 Chr.3: 112116659-112116659 on Build GRCh38
rs778148012 Chr.3: 112116653-112116653 on Build GRCh38
rs1006270981 Chr.3: 112116539-112116539 on Build GRCh38
rs140644894 Chr.3: 112116561-112116565 on Build GRCh38
rs747352242 Chr.3: 112116655-112116655 on Build GRCh38
rs371959386 Chr.3: 112116617-112116617 on Build GRCh38
rs745383938 Chr.3: 112116665-112116665 on Build GRCh38
rs1033322926 Chr.3: 112116553-112116553 on Build GRCh38
rs554992404 Chr.3: 112116579-112116579 on Build GRCh38
rs753019112 Chr.3: 112116645-112116645 on Build GRCh38

View Full Product Details