Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159073293-159073534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIM2
Entrez Gene ID
9447
Gene Name
absent in melanoma 2
Gene Aliases
PYHIN4
UniGene
Hs.733411
Gene Chromosome Location
Chr.1: 159059226-159132351 on Build GRCh38
Transcripts
NM_004833.1 XM_005245616.4 XM_017002848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4023924 c.85C>A p.L29I Chr.1: 159073415-159073415 on Build GRCh38
675998 c.15C>A p.Y5* Chr.1: 159073485-159073485 on Build GRCh38
675999 c.94G>T p.E32* Chr.1: 159073406-159073406 on Build GRCh38
4140892 c.165T>G p.N55K Chr.1: 159073335-159073335 on Build GRCh38
5760912 c.149C>T p.T50I Chr.1: 159073351-159073351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529181365 Chr.1: 159073462-159073462 on Build GRCh38
rs761027006 Chr.1: 159073446-159073446 on Build GRCh38
rs768522855 Chr.1: 159073448-159073448 on Build GRCh38
rs150884694 Chr.1: 159073476-159073476 on Build GRCh38
rs749902483 Chr.1: 159073358-159073358 on Build GRCh38
rs745924959 Chr.1: 159073415-159073415 on Build GRCh38
rs909204337 Chr.1: 159073507-159073507 on Build GRCh38
rs772343677 Chr.1: 159073428-159073428 on Build GRCh38
rs201923123 Chr.1: 159073414-159073414 on Build GRCh38
rs757699983 Chr.1: 159073503-159073503 on Build GRCh38
rs754168382 Chr.1: 159073502-159073502 on Build GRCh38
rs778132585 Chr.1: 159073491-159073491 on Build GRCh38
rs149324922 Chr.1: 159073327-159073327 on Build GRCh38
rs754660736 Chr.1: 159073485-159073485 on Build GRCh38
rs761674723 Chr.1: 159073349-159073349 on Build GRCh38
rs201119668 Chr.1: 159073439-159073439 on Build GRCh38
rs765925699 Chr.1: 159073469-159073469 on Build GRCh38
rs760542046 Chr.1: 159073326-159073326 on Build GRCh38
rs753792783 Chr.1: 159073342-159073342 on Build GRCh38
rs986086448 Chr.1: 159073359-159073359 on Build GRCh38
rs746093551 Chr.1: 159073471-159073471 on Build GRCh38
rs765097282 Chr.1: 159073461-159073461 on Build GRCh38
rs755133172 Chr.1: 159073394-159073394 on Build GRCh38
rs140801792 Chr.1: 159073407-159073407 on Build GRCh38
rs756452296 Chr.1: 159073396-159073396 on Build GRCh38
rs776317433 Chr.1: 159073451-159073451 on Build GRCh38
rs935115122 Chr.1: 159073492-159073492 on Build GRCh38
rs770876017 Chr.1: 159073411-159073414 on Build GRCh38
rs762422070 Chr.1: 159073465-159073465 on Build GRCh38
rs761739654 Chr.1: 159073456-159073456 on Build GRCh38
rs774248430 Chr.1: 159073425-159073425 on Build GRCh38
rs749220389 Chr.1: 159073365-159073366 on Build GRCh38
rs200020470 Chr.1: 159073404-159073404 on Build GRCh38
rs2276405 Chr.1: 159073406-159073406 on Build GRCh38
rs748229238 Chr.1: 159073380-159073380 on Build GRCh38
rs758008523 Chr.1: 159073362-159073362 on Build GRCh38
rs575843618 Chr.1: 159073399-159073399 on Build GRCh38
rs147420667 Chr.1: 159073389-159073389 on Build GRCh38
rs987873854 Chr.1: 159073355-159073355 on Build GRCh38

View Full Product Details