Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210216188-210216441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADL
Entrez Gene ID
33
Gene Name
acyl-CoA dehydrogenase, long chain
Gene Aliases
ACAD4, LCAD
UniGene
Hs.471277
Gene Chromosome Location
Chr.2: 210187990-210225491 on Build GRCh38
Transcripts
NM_001608.3 XM_005246517.4 XM_017003955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1405053 c.487G>A p.A163T Chr.2: 210216396-210216396 on Build GRCh38
719476 c.476C>A p.P159H Chr.2: 210216407-210216407 on Build GRCh38
6188924 c.454G>A p.E152K Chr.2: 210216429-210216429 on Build GRCh38
1631792 c.475C>T p.P159S Chr.2: 210216408-210216408 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780612373 Chr.2: 210216324-210216324 on Build GRCh38
rs747282926 Chr.2: 210216366-210216366 on Build GRCh38
rs140175954 Chr.2: 210216402-210216402 on Build GRCh38
rs149601791 Chr.2: 210216349-210216349 on Build GRCh38
rs755567740 Chr.2: 210216331-210216331 on Build GRCh38
rs763504921 Chr.2: 210216301-210216301 on Build GRCh38
rs781711861 Chr.2: 210216307-210216307 on Build GRCh38
rs995959376 Chr.2: 210216373-210216373 on Build GRCh38
rs953094269 Chr.2: 210216292-210216292 on Build GRCh38
rs779301691 Chr.2: 210216342-210216342 on Build GRCh38
rs748196595 Chr.2: 210216344-210216344 on Build GRCh38
rs769543802 Chr.2: 210216368-210216368 on Build GRCh38
rs756495887 Chr.2: 210216320-210216320 on Build GRCh38
rs949468130 Chr.2: 210216357-210216357 on Build GRCh38
rs997178606 Chr.2: 210216239-210216239 on Build GRCh38
rs767157468 Chr.2: 210216401-210216401 on Build GRCh38
rs761345281 Chr.2: 210216399-210216399 on Build GRCh38
rs777926458 Chr.2: 210216354-210216354 on Build GRCh38
rs750131459 Chr.2: 210216314-210216314 on Build GRCh38
rs1030382394 Chr.2: 210216281-210216281 on Build GRCh38
rs768577146 Chr.2: 210216375-210216375 on Build GRCh38
rs181860201 Chr.2: 210216312-210216312 on Build GRCh38
rs938034442 Chr.2: 210216395-210216395 on Build GRCh38
rs1051507975 Chr.2: 210216377-210216377 on Build GRCh38
rs774160815 Chr.2: 210216386-210216386 on Build GRCh38
rs374042736 Chr.2: 210216305-210216305 on Build GRCh38
rs565750029 Chr.2: 210216369-210216369 on Build GRCh38
rs189433918 Chr.2: 210216311-210216311 on Build GRCh38
rs746942085 Chr.2: 210216299-210216300 on Build GRCh38
rs576648987 Chr.2: 210216225-210216225 on Build GRCh38
rs369881387 Chr.2: 210216296-210216296 on Build GRCh38
rs930126004 Chr.2: 210216297-210216297 on Build GRCh38
rs12104563 Chr.2: 210216326-210216326 on Build GRCh38

View Full Product Details