Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57803108-57803381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVIL
Entrez Gene ID
10677
Gene Name
advillin
Gene Aliases
ADVIL, DOC6, p92
UniGene
Hs.584854
Gene Chromosome Location
Chr.12: 57797372-57818704 on Build GRCh38
Transcripts
NM_006576.3 XM_017018710.1 XM_017018711.1 XM_017018712.1 XM_017018713.1 XM_017018714.1 XM_017018715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5349844 c.1850G>T p.R617L Chr.12: 57803359-57803359 on Build GRCh38
1184265 c.1862G>A p.C621Y Chr.12: 57803347-57803347 on Build GRCh38
942246 c.1870A>G p.K624E Chr.12: 57803339-57803339 on Build GRCh38
4664784 c.1911C>A p.T637T Chr.12: 57803298-57803298 on Build GRCh38
5673497 c.1866C>T p.S622S Chr.12: 57803343-57803343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755234375 Chr.12: 57803241-57803242 on Build GRCh38
rs571352892 Chr.12: 57803189-57803189 on Build GRCh38
rs762132263 Chr.12: 57803282-57803282 on Build GRCh38
rs765836504 Chr.12: 57803324-57803324 on Build GRCh38
rs755758111 Chr.12: 57803229-57803229 on Build GRCh38
rs746790888 Chr.12: 57803201-57803201 on Build GRCh38
rs760892601 Chr.12: 57803269-57803269 on Build GRCh38
rs550362614 Chr.12: 57803183-57803183 on Build GRCh38
rs776411837 Chr.12: 57803274-57803274 on Build GRCh38
rs772035997 Chr.12: 57803257-57803257 on Build GRCh38
rs754916314 Chr.12: 57803246-57803246 on Build GRCh38
rs796190671 Chr.12: 57803181-57803181 on Build GRCh38
rs1048665076 Chr.12: 57803252-57803252 on Build GRCh38
rs770947286 Chr.12: 57803236-57803236 on Build GRCh38
rs777737597 Chr.12: 57803230-57803230 on Build GRCh38
rs774128031 Chr.12: 57803239-57803239 on Build GRCh38
rs1012399728 Chr.12: 57803291-57803291 on Build GRCh38
rs371416960 Chr.12: 57803218-57803218 on Build GRCh38
rs772014206 Chr.12: 57803134-57803134 on Build GRCh38
rs553774286 Chr.12: 57803322-57803322 on Build GRCh38
rs778889853 Chr.12: 57803341-57803341 on Build GRCh38
rs747815367 Chr.12: 57803227-57803227 on Build GRCh38
rs764262075 Chr.12: 57803271-57803271 on Build GRCh38
rs1051111937 Chr.12: 57803160-57803160 on Build GRCh38
rs749040177 Chr.12: 57803231-57803231 on Build GRCh38
rs780954916 Chr.12: 57803223-57803223 on Build GRCh38
rs749137539 Chr.12: 57803353-57803353 on Build GRCh38
rs754440344 Chr.12: 57803219-57803219 on Build GRCh38
rs145662274 Chr.12: 57803264-57803264 on Build GRCh38
rs940343362 Chr.12: 57803171-57803171 on Build GRCh38
rs779240391 Chr.12: 57803217-57803217 on Build GRCh38
rs141510587 Chr.12: 57803335-57803335 on Build GRCh38
rs766937793 Chr.12: 57803333-57803333 on Build GRCh38
rs758880730 Chr.12: 57803325-57803325 on Build GRCh38
rs1046436310 Chr.12: 57803306-57803306 on Build GRCh38

View Full Product Details