Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26906219-26906444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASUN
Entrez Gene ID
55726
Gene Name
asunder, spermatogenesis regulator
Gene Aliases
C12orf11, GCT1, Mat89Bb, NET48, SPATA30
UniGene
Hs.505077
Gene Chromosome Location
Chr.12: 26905171-26938326 on Build GRCh38
Transcripts
NM_018164.2 XM_005253422.3 XM_011520750.2 XM_011520751.2 XM_011520752.2 XM_011520753.2 XM_017019632.1 XM_017019633.1 XM_017019634.1 XM_017019635.1 XM_017019636.1 XM_017019637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
468195 c.2044A>T p.R682* Chr.12: 26906339-26906339 on Build GRCh38
6417465 c.2081G>T p.G694V Chr.12: 26906302-26906302 on Build GRCh38
1605998 c.2051A>C p.E684A Chr.12: 26906332-26906332 on Build GRCh38
3398639 c.1983C>A p.I661I Chr.12: 26906400-26906400 on Build GRCh38
381880 c.2071G>A p.E691K Chr.12: 26906312-26906312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770313037 Chr.12: 26906252-26906253 on Build GRCh38
rs781228627 Chr.12: 26906266-26906266 on Build GRCh38
rs759233692 Chr.12: 26906388-26906388 on Build GRCh38
rs963692758 Chr.12: 26906301-26906301 on Build GRCh38
rs779298735 Chr.12: 26906294-26906294 on Build GRCh38
rs748856884 Chr.12: 26906252-26906256 on Build GRCh38
rs753447309 Chr.12: 26906386-26906386 on Build GRCh38
rs758849418 Chr.12: 26906419-26906419 on Build GRCh38
rs368567088 Chr.12: 26906330-26906330 on Build GRCh38
rs546669638 Chr.12: 26906278-26906278 on Build GRCh38
rs975013767 Chr.12: 26906326-26906326 on Build GRCh38
rs773214357 Chr.12: 26906359-26906359 on Build GRCh38
rs778404449 Chr.12: 26906257-26906257 on Build GRCh38
rs753102255 Chr.12: 26906416-26906416 on Build GRCh38
rs1488052 Chr.12: 26906291-26906291 on Build GRCh38
rs529453556 Chr.12: 26906370-26906370 on Build GRCh38
rs370208549 Chr.12: 26906288-26906288 on Build GRCh38
rs770047393 Chr.12: 26906285-26906285 on Build GRCh38
rs369497577 Chr.12: 26906399-26906399 on Build GRCh38
rs771043579 Chr.12: 26906262-26906262 on Build GRCh38
rs765053308 Chr.12: 26906415-26906415 on Build GRCh38
rs758973230 Chr.12: 26906254-26906254 on Build GRCh38
rs74072633 Chr.12: 26906261-26906261 on Build GRCh38
rs760064652 Chr.12: 26906360-26906360 on Build GRCh38
rs146283660 Chr.12: 26906257-26906258 on Build GRCh38

View Full Product Details