Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56599118-56599391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ2A
Entrez Gene ID
11176
Gene Name
bromodomain adjacent to zinc finger domain 2A
Gene Aliases
TIP5, WALp3
UniGene
Hs.314263
Gene Chromosome Location
Chr.12: 56595596-56636816 on Build GRCh38
Transcripts
NM_001300905.1 NM_013449.3 XM_011537797.2 XM_011537798.2 XM_011537799.2 XM_011537800.2 XM_011537801.2 XM_011537803.2 XM_011537804.2 XM_011537805.2 XM_011537806.2 XM_011537807.2 XM_011537809.2 XM_017018740.1 XM_017018741.1 XM_017018742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6000767 c.5350C>A p.P1784T Chr.12: 56599187-56599187 on Build GRCh38
941725 c.5329C>T p.R1777C Chr.12: 56599316-56599316 on Build GRCh38
4643744 c.5394T>G p.D1798E Chr.12: 56599143-56599143 on Build GRCh38
5492978 c.5333G>A p.G1778D Chr.12: 56599312-56599312 on Build GRCh38
5375642 c.5379G>A p.R1793R Chr.12: 56599158-56599158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773173724 Chr.12: 56599287-56599287 on Build GRCh38
rs901659676 Chr.12: 56599283-56599283 on Build GRCh38
rs763059446 Chr.12: 56599204-56599204 on Build GRCh38
rs538325571 Chr.12: 56599252-56599252 on Build GRCh38
rs748860333 Chr.12: 56599178-56599178 on Build GRCh38
rs768801520 Chr.12: 56599262-56599262 on Build GRCh38
rs757239924 Chr.12: 56599174-56599174 on Build GRCh38
rs757902684 Chr.12: 56599322-56599322 on Build GRCh38
rs767450370 Chr.12: 56599220-56599220 on Build GRCh38
rs761745086 Chr.12: 56599218-56599218 on Build GRCh38
rs757492484 Chr.12: 56599277-56599279 on Build GRCh38
rs962780557 Chr.12: 56599355-56599355 on Build GRCh38
rs760456958 Chr.12: 56599288-56599288 on Build GRCh38
rs746266154 Chr.12: 56599256-56599256 on Build GRCh38
rs1014975877 Chr.12: 56599237-56599237 on Build GRCh38
rs754596264 Chr.12: 56599242-56599242 on Build GRCh38
rs756492383 Chr.12: 56599343-56599343 on Build GRCh38
rs541491077 Chr.12: 56599210-56599210 on Build GRCh38
rs747236357 Chr.12: 56599336-56599336 on Build GRCh38
rs553292948 Chr.12: 56599163-56599163 on Build GRCh38
rs745384480 Chr.12: 56599323-56599323 on Build GRCh38
rs778492865 Chr.12: 56599185-56599185 on Build GRCh38
rs201074705 Chr.12: 56599269-56599269 on Build GRCh38
rs754510339 Chr.12: 56599180-56599180 on Build GRCh38
rs926779460 Chr.12: 56599188-56599188 on Build GRCh38
rs762971471 Chr.12: 56599159-56599159 on Build GRCh38
rs945630741 Chr.12: 56599253-56599253 on Build GRCh38
rs767230458 Chr.12: 56599172-56599172 on Build GRCh38
rs746199531 Chr.12: 56599193-56599193 on Build GRCh38
rs373086268 Chr.12: 56599155-56599155 on Build GRCh38
rs757732182 Chr.12: 56599255-56599255 on Build GRCh38
rs747503106 Chr.12: 56599186-56599186 on Build GRCh38
rs761832887 Chr.12: 56599270-56599270 on Build GRCh38
rs775758301 Chr.12: 56599203-56599203 on Build GRCh38
rs201934985 Chr.12: 56599234-56599234 on Build GRCh38
rs200068250 Chr.12: 56599294-56599294 on Build GRCh38
rs1026122587 Chr.12: 56599296-56599296 on Build GRCh38
rs776860193 Chr.12: 56599192-56599192 on Build GRCh38
rs373446671 Chr.12: 56599303-56599303 on Build GRCh38
rs372009622 Chr.12: 56599173-56599173 on Build GRCh38
rs780403397 Chr.12: 56599353-56599353 on Build GRCh38
rs749527991 Chr.12: 56599261-56599261 on Build GRCh38
rs949760498 Chr.12: 56599312-56599312 on Build GRCh38
rs367731696 Chr.12: 56599148-56599148 on Build GRCh38
rs753539195 Chr.12: 56599239-56599239 on Build GRCh38
rs376855510 Chr.12: 56599314-56599314 on Build GRCh38
rs764067035 Chr.12: 56599160-56599160 on Build GRCh38
rs966039433 Chr.12: 56599238-56599238 on Build GRCh38
rs779712895 Chr.12: 56599177-56599177 on Build GRCh38
rs375310491 Chr.12: 56599211-56599211 on Build GRCh38
rs771409057 Chr.12: 56599191-56599191 on Build GRCh38
rs370265628 Chr.12: 56599340-56599340 on Build GRCh38
rs765991560 Chr.12: 56599235-56599235 on Build GRCh38
rs374233405 Chr.12: 56599258-56599258 on Build GRCh38
rs750208210 Chr.12: 56599227-56599227 on Build GRCh38
rs200742397 Chr.12: 56599171-56599171 on Build GRCh38
rs369538550 Chr.12: 56599246-56599246 on Build GRCh38
rs769930015 Chr.12: 56599197-56599197 on Build GRCh38
rs886329180 Chr.12: 56599315-56599315 on Build GRCh38
rs771956454 Chr.12: 56599284-56599284 on Build GRCh38

View Full Product Details