Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222908720-222908985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL3
Entrez Gene ID
2181
Gene Name
acyl-CoA synthetase long-chain family member 3
Gene Aliases
ACS3, FACL3, PRO2194
UniGene
Hs.655772
Gene Chromosome Location
Chr.2: 222860984-222944624 on Build GRCh38
Transcripts
NM_004457.3 NM_203372.1 XM_017003584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592578 c.135C>T p.S45S Chr.2: 222908907-222908907 on Build GRCh38
4499767 c.54C>G p.I18M Chr.2: 222908826-222908826 on Build GRCh38
7135485 c.161G>T p.R54L Chr.2: 222908933-222908933 on Build GRCh38
5019778 c.33C>T p.T11T Chr.2: 222908805-222908805 on Build GRCh38
5639784 c.12C>T p.H4H Chr.2: 222908784-222908784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764951469 Chr.2: 222908758-222908758 on Build GRCh38
rs748652402 Chr.2: 222908803-222908803 on Build GRCh38
rs984267358 Chr.2: 222908798-222908798 on Build GRCh38
rs146044900 Chr.2: 222908805-222908805 on Build GRCh38
rs556163126 Chr.2: 222908824-222908824 on Build GRCh38
rs756317266 Chr.2: 222908785-222908785 on Build GRCh38
rs745502494 Chr.2: 222908911-222908911 on Build GRCh38
rs758298551 Chr.2: 222908780-222908780 on Build GRCh38
rs755191431 Chr.2: 222908875-222908875 on Build GRCh38
rs749476167 Chr.2: 222908791-222908791 on Build GRCh38
rs768445601 Chr.2: 222908958-222908958 on Build GRCh38
rs367766958 Chr.2: 222908833-222908833 on Build GRCh38
rs542262128 Chr.2: 222908823-222908823 on Build GRCh38
rs750500466 Chr.2: 222908867-222908867 on Build GRCh38
rs368211308 Chr.2: 222908784-222908784 on Build GRCh38
rs928788228 Chr.2: 222908782-222908782 on Build GRCh38
rs755313153 Chr.2: 222908792-222908792 on Build GRCh38
rs770293736 Chr.2: 222908781-222908781 on Build GRCh38
rs777402740 Chr.2: 222908806-222908806 on Build GRCh38
rs749555080 Chr.2: 222908870-222908870 on Build GRCh38
rs759132128 Chr.2: 222908831-222908831 on Build GRCh38
rs754047277 Chr.2: 222908874-222908874 on Build GRCh38
rs758966790 Chr.2: 222908891-222908891 on Build GRCh38
rs142186038 Chr.2: 222908872-222908872 on Build GRCh38
rs762804255 Chr.2: 222908949-222908949 on Build GRCh38
rs762854435 Chr.2: 222908852-222908852 on Build GRCh38
rs370395971 Chr.2: 222908787-222908787 on Build GRCh38
rs373911521 Chr.2: 222908799-222908799 on Build GRCh38
rs1011019757 Chr.2: 222908860-222908860 on Build GRCh38
rs766190690 Chr.2: 222908747-222908747 on Build GRCh38
rs201614126 Chr.2: 222908882-222908882 on Build GRCh38
rs767057399 Chr.2: 222908897-222908897 on Build GRCh38
rs776560167 Chr.2: 222908749-222908749 on Build GRCh38
rs775882417 Chr.2: 222908773-222908798 on Build GRCh38
rs775187847 Chr.2: 222908851-222908851 on Build GRCh38
rs202110843 Chr.2: 222908754-222908754 on Build GRCh38
rs780598298 Chr.2: 222908910-222908910 on Build GRCh38
rs775341174 Chr.2: 222908929-222908929 on Build GRCh38
rs764077633 Chr.2: 222908866-222908866 on Build GRCh38
rs115266165 Chr.2: 222908773-222908773 on Build GRCh38
rs961458463 Chr.2: 222908908-222908908 on Build GRCh38
rs776285728 Chr.2: 222908830-222908830 on Build GRCh38
rs778355721 Chr.2: 222908892-222908892 on Build GRCh38
rs141059382 Chr.2: 222908907-222908907 on Build GRCh38
rs779308602 Chr.2: 222908881-222908881 on Build GRCh38

View Full Product Details