Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72232942-72233173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC16
Entrez Gene ID
119504
Gene Name
anaphase promoting complex subunit 16
Gene Aliases
APC16, C10orf104, CENP-27, MSAG, bA570G20.3
UniGene
Hs.426296
Gene Chromosome Location
Chr.10: 72216000-72235860 on Build GRCh38
Transcripts
NM_001242546.1 NM_001242547.1 NM_001242548.1 NM_173473.3 NR_038391.1 NR_038392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599531 c.327A>G p.S109S Chr.10: 72233110-72233110 on Build GRCh38
1724451 c.283_284insG p.F96fs*2 Chr.10: 72233066-72233067 on Build GRCh38
1683434 c.285_286insG p.F96fs*2 Chr.10: 72233068-72233069 on Build GRCh38
6568558 c.242A>C p.K81T Chr.10: 72233025-72233025 on Build GRCh38
5587168 c.320C>T p.P107L Chr.10: 72233103-72233103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748812488 Chr.10: 72233067-72233067 on Build GRCh38
rs138956935 Chr.10: 72233013-72233013 on Build GRCh38
rs755407083 Chr.10: 72233059-72233059 on Build GRCh38
rs769339108 Chr.10: 72232985-72232985 on Build GRCh38
rs748761104 Chr.10: 72233130-72233130 on Build GRCh38
rs373225911 Chr.10: 72233060-72233060 on Build GRCh38
rs775278371 Chr.10: 72233081-72233081 on Build GRCh38
rs374220486 Chr.10: 72233119-72233119 on Build GRCh38
rs777225132 Chr.10: 72232965-72232965 on Build GRCh38
rs762664254 Chr.10: 72232991-72232991 on Build GRCh38
rs201778398 Chr.10: 72233143-72233143 on Build GRCh38
rs370996491 Chr.10: 72233142-72233142 on Build GRCh38
rs374611476 Chr.10: 72232964-72232964 on Build GRCh38
rs750534184 Chr.10: 72233047-72233047 on Build GRCh38
rs770515817 Chr.10: 72233137-72233137 on Build GRCh38
rs751852740 Chr.10: 72233118-72233118 on Build GRCh38
rs772645649 Chr.10: 72233065-72233065 on Build GRCh38
rs927789792 Chr.10: 72233085-72233085 on Build GRCh38
rs9275 Chr.10: 72233104-72233104 on Build GRCh38
rs758581509 Chr.10: 72233112-72233112 on Build GRCh38
rs145808346 Chr.10: 72233113-72233113 on Build GRCh38
rs769286255 Chr.10: 72233062-72233062 on Build GRCh38
rs956393732 Chr.10: 72232978-72232978 on Build GRCh38
rs930627472 Chr.10: 72233117-72233117 on Build GRCh38
rs778278608 Chr.10: 72233127-72233127 on Build GRCh38
rs756558579 Chr.10: 72233124-72233124 on Build GRCh38
rs150188116 Chr.10: 72233012-72233012 on Build GRCh38
rs780345444 Chr.10: 72233053-72233053 on Build GRCh38
rs780751422 Chr.10: 72233026-72233027 on Build GRCh38
rs770445774 Chr.10: 72233069-72233069 on Build GRCh38
rs1033463145 Chr.10: 72233066-72233066 on Build GRCh38
rs767395303 Chr.10: 72233080-72233080 on Build GRCh38
rs770299440 Chr.10: 72232982-72232982 on Build GRCh38
rs752721096 Chr.10: 72233015-72233015 on Build GRCh38
rs200000965 Chr.10: 72233016-72233016 on Build GRCh38
rs774088740 Chr.10: 72233077-72233077 on Build GRCh38
rs147406330 Chr.10: 72233066-72233067 on Build GRCh38
rs774575440 Chr.10: 72233154-72233155 on Build GRCh38
rs759220497 Chr.10: 72233078-72233078 on Build GRCh38
rs13980 Chr.10: 72233121-72233121 on Build GRCh38
rs764815399 Chr.10: 72232990-72232990 on Build GRCh38
rs773962859 Chr.10: 72233008-72233008 on Build GRCh38
rs376962158 Chr.10: 72232994-72232994 on Build GRCh38
rs1040509527 Chr.10: 72233151-72233151 on Build GRCh38
rs749831431 Chr.10: 72233086-72233086 on Build GRCh38
rs758634664 Chr.10: 72233048-72233048 on Build GRCh38
rs200589401 Chr.10: 72233144-72233144 on Build GRCh38
rs747576369 Chr.10: 72232973-72232973 on Build GRCh38
rs11553567 Chr.10: 72233091-72233091 on Build GRCh38
rs747296126 Chr.10: 72233055-72233055 on Build GRCh38
rs765472984 Chr.10: 72233032-72233032 on Build GRCh38
rs768813451 Chr.10: 72233142-72233155 on Build GRCh38
rs566040344 Chr.10: 72233100-72233100 on Build GRCh38
rs764137906 Chr.10: 72233103-72233103 on Build GRCh38

View Full Product Details