Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97913356-97913603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMO
Entrez Gene ID
51531
Gene Name
tRNA methyltransferase O
Gene Aliases
C9orf156, HSPC219, NAP1
UniGene
Hs.9196
Gene Chromosome Location
Chr.9: 97876321-97922570 on Build GRCh38
Transcripts
NM_016481.4 XM_005252047.3 XM_005252048.4 XM_005252049.4 XM_005252050.4 XM_011518776.2 XM_011518777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616848 c.250A>G p.I84V Chr.9: 97913557-97913557 on Build GRCh38
4668091 c.289T>G p.C97G Chr.9: 97913521-97913521 on Build GRCh38
7224968 c.309G>A p.R103R Chr.9: 97913495-97913495 on Build GRCh38
3432689 c.394A>C p.K132Q Chr.9: 97913410-97913410 on Build GRCh38
3432690 c.397A>C p.K133Q Chr.9: 97913410-97913410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201918701 Chr.9: 97913535-97913535 on Build GRCh38
rs769631848 Chr.9: 97913408-97913408 on Build GRCh38
rs1000279633 Chr.9: 97913496-97913496 on Build GRCh38
rs763003299 Chr.9: 97913412-97913412 on Build GRCh38
rs747862063 Chr.9: 97913501-97913501 on Build GRCh38
rs776472704 Chr.9: 97913394-97913394 on Build GRCh38
rs765666808 Chr.9: 97913449-97913449 on Build GRCh38
rs754104802 Chr.9: 97913444-97913444 on Build GRCh38
rs370421951 Chr.9: 97913389-97913389 on Build GRCh38
rs74477697 Chr.9: 97913541-97913541 on Build GRCh38
rs373097511 Chr.9: 97913522-97913522 on Build GRCh38
rs764331340 Chr.9: 97913565-97913569 on Build GRCh38
rs757732466 Chr.9: 97913532-97913532 on Build GRCh38
rs767685242 Chr.9: 97913426-97913426 on Build GRCh38
rs77668890 Chr.9: 97913495-97913495 on Build GRCh38
rs764238494 Chr.9: 97913562-97913562 on Build GRCh38
rs763111090 Chr.9: 97913476-97913477 on Build GRCh38
rs779533593 Chr.9: 97913471-97913471 on Build GRCh38
rs775755150 Chr.9: 97913434-97913434 on Build GRCh38
rs746580178 Chr.9: 97913481-97913481 on Build GRCh38
rs929123807 Chr.9: 97913388-97913388 on Build GRCh38
rs758942922 Chr.9: 97913464-97913464 on Build GRCh38
rs1044165570 Chr.9: 97913485-97913485 on Build GRCh38
rs750882392 Chr.9: 97913451-97913451 on Build GRCh38
rs759854012 Chr.9: 97913391-97913391 on Build GRCh38
rs141236958 Chr.9: 97913442-97913442 on Build GRCh38
rs760779497 Chr.9: 97913543-97913543 on Build GRCh38
rs754529337 Chr.9: 97913487-97913487 on Build GRCh38
rs1127703 Chr.9: 97913534-97913534 on Build GRCh38
rs760978487 Chr.9: 97913437-97913437 on Build GRCh38
rs747835480 Chr.9: 97913397-97913397 on Build GRCh38
rs751819442 Chr.9: 97913564-97913565 on Build GRCh38
rs375908838 Chr.9: 97913386-97913386 on Build GRCh38
rs766127779 Chr.9: 97913459-97913459 on Build GRCh38
rs757496935 Chr.9: 97913448-97913448 on Build GRCh38
rs747606470 Chr.9: 97913394-97913394 on Build GRCh38
rs753587623 Chr.9: 97913464-97913465 on Build GRCh38
rs773106000 Chr.9: 97913530-97913530 on Build GRCh38

View Full Product Details