Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 78955142-78955405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSB
Entrez Gene ID
411
Gene Name
arylsulfatase B
Gene Aliases
ASB, G4S, MPS6
UniGene
Hs.149103
Gene Chromosome Location
Chr.5: 78777209-78986534 on Build GRCh38
Transcripts
NM_000046.3 NM_198709.2 XM_011543390.1 XM_011543391.2 XM_011543392.2 XM_011543393.1 XM_017009471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4869959 c.875A>G p.N292S Chr.5: 78955318-78955318 on Build GRCh38
1183680 c.865C>A p.L289I Chr.5: 78955328-78955328 on Build GRCh38
6171950 c.857G>A p.S286N Chr.5: 78955336-78955336 on Build GRCh38
3618023 c.813C>T p.S271S Chr.5: 78955380-78955380 on Build GRCh38
3618022 c.813C>T p.S271S Chr.5: 78955380-78955380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191984227 Chr.5: 78955239-78955239 on Build GRCh38
rs765506970 Chr.5: 78955315-78955315 on Build GRCh38
rs760136403 Chr.5: 78955279-78955279 on Build GRCh38
rs781043229 Chr.5: 78955384-78955384 on Build GRCh38
rs753051996 Chr.5: 78955325-78955325 on Build GRCh38
rs758841265 Chr.5: 78955328-78955328 on Build GRCh38
rs573613390 Chr.5: 78955288-78955288 on Build GRCh38
rs751323091 Chr.5: 78955273-78955273 on Build GRCh38
rs778728769 Chr.5: 78955381-78955381 on Build GRCh38
rs755579897 Chr.5: 78955314-78955314 on Build GRCh38
rs768696049 Chr.5: 78955375-78955375 on Build GRCh38
rs373376330 Chr.5: 78955249-78955249 on Build GRCh38
rs747255542 Chr.5: 78955352-78955352 on Build GRCh38
rs536856219 Chr.5: 78955175-78955175 on Build GRCh38
rs757019634 Chr.5: 78955380-78955380 on Build GRCh38
rs545693472 Chr.5: 78955334-78955334 on Build GRCh38
rs774268494 Chr.5: 78955250-78955250 on Build GRCh38
rs767095787 Chr.5: 78955271-78955271 on Build GRCh38
rs963988879 Chr.5: 78955245-78955245 on Build GRCh38
rs752398894 Chr.5: 78955221-78955221 on Build GRCh38
rs763681766 Chr.5: 78955212-78955212 on Build GRCh38
rs765925648 Chr.5: 78955281-78955281 on Build GRCh38
rs975766834 Chr.5: 78955247-78955247 on Build GRCh38
rs376522527 Chr.5: 78955371-78955371 on Build GRCh38
rs761353977 Chr.5: 78955259-78955259 on Build GRCh38
rs942175743 Chr.5: 78955313-78955313 on Build GRCh38
rs757387778 Chr.5: 78955360-78955360 on Build GRCh38
rs34555226 Chr.5: 78955341-78955341 on Build GRCh38
rs755676638 Chr.5: 78955226-78955226 on Build GRCh38
rs781369253 Chr.5: 78955367-78955367 on Build GRCh38
rs1005512928 Chr.5: 78955222-78955222 on Build GRCh38

View Full Product Details