Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66851434-66851705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA7
Entrez Gene ID
766
Gene Name
carbonic anhydrase 7
Gene Aliases
CAVII
UniGene
Hs.37014
Gene Chromosome Location
Chr.16: 66844379-66854149 on Build GRCh38
Transcripts
NM_001014435.1 NM_005182.2 XM_005256136.3 XM_011523311.1 XM_011523312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997332 c.449delT p.L150fs*10 Chr.16: 66851554-66851554 on Build GRCh38
5851185 c.400G>A p.G134R Chr.16: 66851505-66851505 on Build GRCh38
5669040 c.461A>G p.D154G Chr.16: 66851671-66851671 on Build GRCh38
6617126 c.408G>A p.A136A Chr.16: 66851513-66851513 on Build GRCh38
1493712 c.378T>A p.N126K Chr.16: 66851483-66851483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764659485 Chr.16: 66851598-66851598 on Build GRCh38
rs905743539 Chr.16: 66851536-66851536 on Build GRCh38
rs764520768 Chr.16: 66851506-66851506 on Build GRCh38
rs775864644 Chr.16: 66851589-66851589 on Build GRCh38
rs781534179 Chr.16: 66851516-66851516 on Build GRCh38
rs775071324 Chr.16: 66851673-66851673 on Build GRCh38
rs74938147 Chr.16: 66851627-66851627 on Build GRCh38
rs756342313 Chr.16: 66851625-66851625 on Build GRCh38
rs759866965 Chr.16: 66851455-66851455 on Build GRCh38
rs774762898 Chr.16: 66851600-66851600 on Build GRCh38
rs771991681 Chr.16: 66851555-66851555 on Build GRCh38
rs777636101 Chr.16: 66851561-66851561 on Build GRCh38
rs759147522 Chr.16: 66851580-66851580 on Build GRCh38
rs750724409 Chr.16: 66851519-66851519 on Build GRCh38
rs781736939 Chr.16: 66851614-66851615 on Build GRCh38
rs376555362 Chr.16: 66851457-66851457 on Build GRCh38
rs76870487 Chr.16: 66851579-66851579 on Build GRCh38
rs777741146 Chr.16: 66851640-66851640 on Build GRCh38
rs758163789 Chr.16: 66851636-66851636 on Build GRCh38
rs1004649889 Chr.16: 66851663-66851663 on Build GRCh38
rs757348553 Chr.16: 66851514-66851514 on Build GRCh38
rs757214019 Chr.16: 66851655-66851655 on Build GRCh38
rs747902788 Chr.16: 66851549-66851549 on Build GRCh38
rs746868763 Chr.16: 66851650-66851650 on Build GRCh38
rs753033289 Chr.16: 66851471-66851471 on Build GRCh38
rs770473978 Chr.16: 66851572-66851572 on Build GRCh38
rs372793869 Chr.16: 66851488-66851488 on Build GRCh38
rs201896800 Chr.16: 66851672-66851672 on Build GRCh38
rs768952080 Chr.16: 66851684-66851684 on Build GRCh38
rs116420759 Chr.16: 66851513-66851513 on Build GRCh38
rs1049178765 Chr.16: 66851583-66851583 on Build GRCh38
rs745395739 Chr.16: 66851667-66851667 on Build GRCh38
rs763283489 Chr.16: 66851594-66851594 on Build GRCh38
rs887552280 Chr.16: 66851644-66851644 on Build GRCh38
rs746494062 Chr.16: 66851565-66851565 on Build GRCh38
rs912217770 Chr.16: 66851484-66851484 on Build GRCh38
rs749635386 Chr.16: 66851682-66851682 on Build GRCh38
rs370140698 Chr.16: 66851603-66851603 on Build GRCh38
rs373451178 Chr.16: 66851624-66851624 on Build GRCh38
rs375798363 Chr.16: 66851582-66851582 on Build GRCh38
rs985156841 Chr.16: 66851463-66851463 on Build GRCh38
rs780613658 Chr.16: 66851660-66851660 on Build GRCh38
rs756430457 Chr.16: 66851538-66851538 on Build GRCh38
rs778815563 Chr.16: 66851540-66851540 on Build GRCh38
rs767656521 Chr.16: 66851622-66851622 on Build GRCh38
rs761961593 Chr.16: 66851604-66851604 on Build GRCh38

View Full Product Details