Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42814477-42814730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BECN1
Entrez Gene ID
8678
Gene Name
beclin 1
Gene Aliases
ATG6, VPS30, beclin1
UniGene
Hs.716464
Gene Chromosome Location
Chr.17: 42810132-42824316 on Build GRCh38
Transcripts
NM_001313998.1 NM_001313999.1 NM_001314000.1 NM_003766.4 XM_005257759.2 XM_005257760.3 XM_011525421.2 XM_017025262.1 XM_017025263.1 XM_017025264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4066687 c.962T>C p.M321T Chr.17: 42814542-42814542 on Build GRCh38
6053815 c.969G>T p.L323L Chr.17: 42814535-42814535 on Build GRCh38
7428555 c.933G>T p.V311V Chr.17: 42814571-42814571 on Build GRCh38
706257 c.927G>C p.Q309H Chr.17: 42814577-42814577 on Build GRCh38
4066688 c.859A>G p.N287D Chr.17: 42814645-42814645 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs979013935 Chr.17: 42814540-42814540 on Build GRCh38
rs757626703 Chr.17: 42814612-42814612 on Build GRCh38
rs778217364 Chr.17: 42814600-42814600 on Build GRCh38
rs200971735 Chr.17: 42814588-42814588 on Build GRCh38
rs138015339 Chr.17: 42814696-42814696 on Build GRCh38
rs747398485 Chr.17: 42814611-42814611 on Build GRCh38
rs781351865 Chr.17: 42814616-42814616 on Build GRCh38
rs761641647 Chr.17: 42814548-42814548 on Build GRCh38
rs775164058 Chr.17: 42814630-42814630 on Build GRCh38
rs766581104 Chr.17: 42814555-42814555 on Build GRCh38
rs768754246 Chr.17: 42814654-42814654 on Build GRCh38
rs147341724 Chr.17: 42814514-42814514 on Build GRCh38
rs149225760 Chr.17: 42814526-42814526 on Build GRCh38
rs769533035 Chr.17: 42814629-42814629 on Build GRCh38
rs1017228948 Chr.17: 42814628-42814628 on Build GRCh38
rs747373422 Chr.17: 42814507-42814507 on Build GRCh38
rs754080310 Chr.17: 42814561-42814561 on Build GRCh38
rs752721484 Chr.17: 42814589-42814589 on Build GRCh38
rs200302913 Chr.17: 42814510-42814510 on Build GRCh38
rs758718841 Chr.17: 42814627-42814627 on Build GRCh38
rs747127691 Chr.17: 42814696-42814696 on Build GRCh38
rs922330983 Chr.17: 42814690-42814690 on Build GRCh38
rs1005470436 Chr.17: 42814613-42814613 on Build GRCh38
rs769440747 Chr.17: 42814517-42814517 on Build GRCh38
rs758949080 Chr.17: 42814598-42814598 on Build GRCh38
rs774338036 Chr.17: 42814536-42814536 on Build GRCh38
rs745537306 Chr.17: 42814621-42814621 on Build GRCh38
rs765850038 Chr.17: 42814576-42814576 on Build GRCh38
rs780081389 Chr.17: 42814695-42814696 on Build GRCh38
rs755099766 Chr.17: 42814563-42814563 on Build GRCh38
rs775076091 Chr.17: 42814525-42814525 on Build GRCh38
rs764074580 Chr.17: 42814532-42814532 on Build GRCh38
rs778129564 Chr.17: 42814503-42814503 on Build GRCh38
rs761574649 Chr.17: 42814685-42814685 on Build GRCh38
rs774345875 Chr.17: 42814684-42814684 on Build GRCh38
rs748786575 Chr.17: 42814651-42814651 on Build GRCh38
rs57695719 Chr.17: 42814515-42814515 on Build GRCh38

View Full Product Details