Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69083173-69083430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA6
Entrez Gene ID
23460
Gene Name
ATP binding cassette subfamily A member 6
Gene Aliases
EST155051
UniGene
Hs.709514
Gene Chromosome Location
Chr.17: 69062044-69142188 on Build GRCh38
Transcripts
NM_080284.2 XM_011524574.2 XM_017024404.1 XM_017024405.1 XM_017024406.1 XM_017024407.1 XM_017024408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147470 c.4465G>T p.G1489* Chr.17: 69083222-69083222 on Build GRCh38
1163170 c.4444C>T p.R1482C Chr.17: 69083243-69083243 on Build GRCh38
4421364 c.4442_4449delACCGTGTG p.D1481fs*9 Chr.17: 69083238-69083245 on Build GRCh38
1563686 c.4461G>T p.V1487V Chr.17: 69083226-69083226 on Build GRCh38
1303261 c.4387G>C p.E1463Q Chr.17: 69083300-69083300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753033213 Chr.17: 69083321-69083321 on Build GRCh38
rs780755597 Chr.17: 69083207-69083207 on Build GRCh38
rs201717023 Chr.17: 69083237-69083237 on Build GRCh38
rs1041247672 Chr.17: 69083322-69083322 on Build GRCh38
rs750069056 Chr.17: 69083286-69083286 on Build GRCh38
rs764820975 Chr.17: 69083283-69083283 on Build GRCh38
rs866757538 Chr.17: 69083349-69083349 on Build GRCh38
rs769637483 Chr.17: 69083382-69083382 on Build GRCh38
rs781467942 Chr.17: 69083384-69083384 on Build GRCh38
rs937839725 Chr.17: 69083343-69083343 on Build GRCh38
rs34865180 Chr.17: 69083343-69083343 on Build GRCh38
rs752169862 Chr.17: 69083312-69083312 on Build GRCh38
rs117323775 Chr.17: 69083254-69083254 on Build GRCh38
rs200105664 Chr.17: 69083242-69083242 on Build GRCh38
rs893715898 Chr.17: 69083397-69083397 on Build GRCh38
rs768462821 Chr.17: 69083255-69083256 on Build GRCh38
rs34498359 Chr.17: 69083313-69083313 on Build GRCh38
rs997025276 Chr.17: 69083347-69083347 on Build GRCh38
rs776297059 Chr.17: 69083275-69083275 on Build GRCh38
rs373967546 Chr.17: 69083203-69083203 on Build GRCh38
rs953544733 Chr.17: 69083208-69083208 on Build GRCh38
rs769911490 Chr.17: 69083241-69083241 on Build GRCh38
rs768509031 Chr.17: 69083351-69083351 on Build GRCh38
rs764039055 Chr.17: 69083271-69083271 on Build GRCh38
rs143326198 Chr.17: 69083300-69083300 on Build GRCh38
rs781396888 Chr.17: 69083231-69083231 on Build GRCh38
rs778981610 Chr.17: 69083338-69083338 on Build GRCh38
rs755459989 Chr.17: 69083220-69083220 on Build GRCh38
rs746948732 Chr.17: 69083219-69083219 on Build GRCh38
rs776613342 Chr.17: 69083294-69083295 on Build GRCh38
rs749934900 Chr.17: 69083198-69083198 on Build GRCh38
rs776584099 Chr.17: 69083380-69083380 on Build GRCh38
rs756449112 Chr.17: 69083327-69083327 on Build GRCh38
rs760596501 Chr.17: 69083260-69083260 on Build GRCh38
rs181258241 Chr.17: 69083243-69083243 on Build GRCh38
rs758022121 Chr.17: 69083298-69083298 on Build GRCh38
rs747788718 Chr.17: 69083301-69083302 on Build GRCh38
rs745885724 Chr.17: 69083344-69083344 on Build GRCh38
rs749477068 Chr.17: 69083336-69083336 on Build GRCh38
rs1015456022 Chr.17: 69083199-69083199 on Build GRCh38
rs878981507 Chr.17: 69083216-69083216 on Build GRCh38
rs201702880 Chr.17: 69083259-69083259 on Build GRCh38
rs758471024 Chr.17: 69083197-69083197 on Build GRCh38
rs765984300 Chr.17: 69083299-69083299 on Build GRCh38
rs761631419 Chr.17: 69083277-69083277 on Build GRCh38
rs149614799 Chr.17: 69083211-69083211 on Build GRCh38
rs771062529 Chr.17: 69083337-69083337 on Build GRCh38
rs992622356 Chr.17: 69083204-69083204 on Build GRCh38
rs773001201 Chr.17: 69083381-69083382 on Build GRCh38
rs781673805 Chr.17: 69083318-69083318 on Build GRCh38

View Full Product Details