Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82449199-82449426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf62
Entrez Gene ID
79415
Gene Name
chromosome 17 open reading frame 62
Gene Aliases
-
UniGene
Hs.163113
Gene Chromosome Location
Chr.17: 82442586-82450831 on Build GRCh38
Transcripts
NM_001033046.3 NM_001100407.2 NM_001100408.2 NM_001193653.1 NM_001193654.1 NM_001193655.1 NM_001193657.1 NR_036514.1 NR_036516.1 NR_036517.1 NR_036518.1 XM_006722293.2 XM_011523606.2 XM_017025072.1 XM_017025073.1 XM_017025074.1 XM_017025075.1 XM_017025076.1 XM_017025077.1 XM_017025078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970447 c.43C>G p.L15V Chr.17: 82449212-82449212 on Build GRCh38
3523866 c.26C>T p.T9I Chr.17: 82449229-82449229 on Build GRCh38
6568079 c.43C>T p.L15L Chr.17: 82449212-82449212 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754033409 Chr.17: 82449289-82449289 on Build GRCh38
rs530198980 Chr.17: 82449383-82449383 on Build GRCh38
rs752876190 Chr.17: 82449232-82449232 on Build GRCh38
rs548495660 Chr.17: 82449384-82449384 on Build GRCh38
rs960363648 Chr.17: 82449326-82449326 on Build GRCh38
rs970106041 Chr.17: 82449262-82449262 on Build GRCh38
rs1050706182 Chr.17: 82449247-82449247 on Build GRCh38
rs377494603 Chr.17: 82449304-82449304 on Build GRCh38
rs373856077 Chr.17: 82449354-82449354 on Build GRCh38
rs746154180 Chr.17: 82449265-82449265 on Build GRCh38
rs760345209 Chr.17: 82449303-82449303 on Build GRCh38
rs111962223 Chr.17: 82449343-82449343 on Build GRCh38
rs771543342 Chr.17: 82449293-82449293 on Build GRCh38
rs775848800 Chr.17: 82449290-82449290 on Build GRCh38
rs1035790184 Chr.17: 82449295-82449295 on Build GRCh38
rs1011712349 Chr.17: 82449307-82449307 on Build GRCh38
rs778180128 Chr.17: 82449249-82449249 on Build GRCh38
rs908412449 Chr.17: 82449349-82449349 on Build GRCh38
rs562977049 Chr.17: 82449360-82449360 on Build GRCh38
rs892050035 Chr.17: 82449234-82449234 on Build GRCh38
rs772902922 Chr.17: 82449294-82449294 on Build GRCh38
rs373528172 Chr.17: 82449228-82449228 on Build GRCh38
rs761383420 Chr.17: 82449291-82449291 on Build GRCh38
rs919825049 Chr.17: 82449381-82449381 on Build GRCh38
rs974017882 Chr.17: 82449375-82449375 on Build GRCh38
rs1011817399 Chr.17: 82449254-82449254 on Build GRCh38
rs142996233 Chr.17: 82449288-82449288 on Build GRCh38
rs754945284 Chr.17: 82449220-82449220 on Build GRCh38
rs755941401 Chr.17: 82449287-82449288 on Build GRCh38
rs747210616 Chr.17: 82449256-82449256 on Build GRCh38
rs113748563 Chr.17: 82449258-82449258 on Build GRCh38
rs375800523 Chr.17: 82449233-82449233 on Build GRCh38

View Full Product Details