Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7983383-7983608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM2
Entrez Gene ID
84448
Gene Name
actin binding LIM protein family member 2
Gene Aliases
-
UniGene
Hs.233404
Gene Chromosome Location
Chr.4: 7965310-8158832 on Build GRCh38
Transcripts
NM_001130083.1 NM_001130084.1 NM_001130085.1 NM_001130086.1 NM_001130087.1 NM_001286688.1 NM_032432.4 XM_005248014.4 XM_005248017.4 XM_005248018.4 XM_005248019.4 XM_005248020.4 XM_005248021.4 XM_005248022.4 XM_005248023.4 XM_005248024.4 XM_005248027.4 XM_005248028.4 XM_005248029.4 XM_005248030.4 XM_006713923.3 XM_006713924.3 XM_006713925.3 XM_006713927.3 XM_011513586.2 XM_017008706.1 XM_017008707.1 XM_017008708.1 XM_017008709.1 XM_017008710.1 XM_017008711.1 XM_017008712.1 XM_017008713.1 XM_017008714.1 XM_017008715.1 XM_017008716.1 XM_017008717.1 XM_017008718.1 XM_017008719.1 XM_017008720.1 XM_017008721.1 XM_017008722.1 XM_017008724.1 XM_017008726.1 XM_017008727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7259956 c.1394-6_1394-5insCT p.? Chr.4: 7983559-7983560 on Build GRCh38
7259955 c.1736-6_1736-5insCT p.? Chr.4: 7983559-7983560 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371079889 Chr.4: 7983438-7983438 on Build GRCh38
rs746566402 Chr.4: 7983573-7983573 on Build GRCh38
rs756409784 Chr.4: 7983560-7983563 on Build GRCh38
rs779460847 Chr.4: 7983572-7983572 on Build GRCh38
rs998548162 Chr.4: 7983495-7983495 on Build GRCh38
rs866670061 Chr.4: 7983518-7983518 on Build GRCh38
rs749446854 Chr.4: 7983511-7983511 on Build GRCh38
rs370012949 Chr.4: 7983569-7983569 on Build GRCh38
rs532562201 Chr.4: 7983509-7983509 on Build GRCh38
rs369888216 Chr.4: 7983476-7983476 on Build GRCh38
rs770773180 Chr.4: 7983581-7983581 on Build GRCh38
rs925495021 Chr.4: 7983422-7983422 on Build GRCh38
rs776417134 Chr.4: 7983515-7983515 on Build GRCh38
rs369579061 Chr.4: 7983526-7983526 on Build GRCh38
rs199606489 Chr.4: 7983517-7983517 on Build GRCh38
rs759444010 Chr.4: 7983516-7983516 on Build GRCh38
rs746302043 Chr.4: 7983506-7983506 on Build GRCh38
rs145815093 Chr.4: 7983439-7983439 on Build GRCh38
rs760531559 Chr.4: 7983514-7983514 on Build GRCh38
rs200961918 Chr.4: 7983559-7983560 on Build GRCh38
rs1052551908 Chr.4: 7983459-7983459 on Build GRCh38
rs948019131 Chr.4: 7983418-7983418 on Build GRCh38
rs776029845 Chr.4: 7983510-7983510 on Build GRCh38
rs772599189 Chr.4: 7983512-7983512 on Build GRCh38
rs373344302 Chr.4: 7983535-7983535 on Build GRCh38
rs1021419398 Chr.4: 7983449-7983449 on Build GRCh38
rs749597966 Chr.4: 7983549-7983549 on Build GRCh38
rs757326290 Chr.4: 7983534-7983534 on Build GRCh38
rs763757729 Chr.4: 7983570-7983571 on Build GRCh38
rs376436867 Chr.4: 7983540-7983540 on Build GRCh38
rs1043749952 Chr.4: 7983420-7983420 on Build GRCh38
rs752913991 Chr.4: 7983560-7983561 on Build GRCh38
rs751650705 Chr.4: 7983533-7983533 on Build GRCh38
rs780423693 Chr.4: 7983541-7983541 on Build GRCh38
rs891319584 Chr.4: 7983464-7983464 on Build GRCh38
rs752497933 Chr.4: 7983525-7983525 on Build GRCh38

View Full Product Details