Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140749228-140749481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAF
Entrez Gene ID
673
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
B-RAF1, B-raf, BRAF1, NS7, RAFB1
UniGene
Hs.550061
Gene Chromosome Location
Chr.7: 140719331-140924764 on Build GRCh38
Transcripts
NM_004333.4 XM_005250045.2 XM_017012558.1 XM_017012559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7081905 c.1983C>T p.N661N Chr.7: 140749296-140749296 on Build GRCh38
6582582 c.1885G>T p.D629Y Chr.7: 140749394-140749394 on Build GRCh38
745348 c.1861-2A>G p.? Chr.7: 140749420-140749420 on Build GRCh38
745349 c.1915G>A p.V639I Chr.7: 140749364-140749364 on Build GRCh38
6475305 c.1977C>A p.I659I Chr.7: 140749302-140749302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766089848 Chr.7: 140749272-140749272 on Build GRCh38
rs369889467 Chr.7: 140749436-140749436 on Build GRCh38
rs1043753843 Chr.7: 140749425-140749425 on Build GRCh38
rs778554187 Chr.7: 140749290-140749290 on Build GRCh38
rs766291057 Chr.7: 140749449-140749449 on Build GRCh38
rs754481830 Chr.7: 140749289-140749289 on Build GRCh38
rs771854433 Chr.7: 140749440-140749440 on Build GRCh38
rs3789806 Chr.7: 140749271-140749271 on Build GRCh38
rs746010267 Chr.7: 140749382-140749382 on Build GRCh38
rs758803218 Chr.7: 140749343-140749343 on Build GRCh38
rs771285548 Chr.7: 140749404-140749404 on Build GRCh38
rs775514667 Chr.7: 140749433-140749433 on Build GRCh38
rs9648696 Chr.7: 140749350-140749350 on Build GRCh38
rs753545284 Chr.7: 140749279-140749279 on Build GRCh38
rs781330698 Chr.7: 140749409-140749409 on Build GRCh38
rs746074624 Chr.7: 140749415-140749415 on Build GRCh38
rs180177042 Chr.7: 140749365-140749365 on Build GRCh38
rs778397955 Chr.7: 140749346-140749346 on Build GRCh38
rs770065462 Chr.7: 140749428-140749428 on Build GRCh38
rs767528258 Chr.7: 140749263-140749263 on Build GRCh38
rs773232782 Chr.7: 140749442-140749442 on Build GRCh38
rs753182578 Chr.7: 140749260-140749260 on Build GRCh38
rs772112410 Chr.7: 140749305-140749305 on Build GRCh38
rs763353075 Chr.7: 140749295-140749297 on Build GRCh38
rs752322193 Chr.7: 140749304-140749304 on Build GRCh38
rs764435055 Chr.7: 140749446-140749450 on Build GRCh38
rs760320159 Chr.7: 140749444-140749444 on Build GRCh38
rs879255380 Chr.7: 140749329-140749329 on Build GRCh38
rs727502905 Chr.7: 140749319-140749319 on Build GRCh38

View Full Product Details