Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110018950-110019211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13A
Entrez Gene ID
88455
Gene Name
ankyrin repeat domain 13A
Gene Aliases
ANKRD13, NY-REN-25
UniGene
Hs.528703
Gene Chromosome Location
Chr.12: 109999169-110039432 on Build GRCh38
Transcripts
NM_033121.1 XM_005253980.2 XM_005253981.3 XM_005253982.3 XM_005253984.1 XM_011538937.1 XM_011538938.2 XM_017020157.1 XM_017020158.1 XM_017020159.1 XM_017020160.1 XM_017020161.1 XM_017020162.1 XM_017020163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1946843 c.606C>T p.F202F Chr.12: 110019200-110019200 on Build GRCh38
5913321 c.574C>T p.H192Y Chr.12: 110019168-110019168 on Build GRCh38
4661754 c.554C>T p.A185V Chr.12: 110019148-110019148 on Build GRCh38
6000823 c.601C>T p.R201C Chr.12: 110019195-110019195 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780258923 Chr.12: 110019142-110019142 on Build GRCh38
rs575695821 Chr.12: 110019073-110019073 on Build GRCh38
rs367745440 Chr.12: 110019169-110019169 on Build GRCh38
rs370743235 Chr.12: 110019109-110019109 on Build GRCh38
rs767285571 Chr.12: 110019106-110019106 on Build GRCh38
rs373926911 Chr.12: 110019151-110019151 on Build GRCh38
rs145153429 Chr.12: 110019149-110019149 on Build GRCh38
rs754667334 Chr.12: 110019024-110019024 on Build GRCh38
rs1008896576 Chr.12: 110019013-110019013 on Build GRCh38
rs759452049 Chr.12: 110019104-110019104 on Build GRCh38
rs890462278 Chr.12: 110018982-110018982 on Build GRCh38
rs371318947 Chr.12: 110019191-110019191 on Build GRCh38
rs748914527 Chr.12: 110019190-110019190 on Build GRCh38
rs772729715 Chr.12: 110019092-110019092 on Build GRCh38
rs747749163 Chr.12: 110019117-110019117 on Build GRCh38
rs944157468 Chr.12: 110019014-110019014 on Build GRCh38
rs866470858 Chr.12: 110019150-110019150 on Build GRCh38
rs368273015 Chr.12: 110019093-110019093 on Build GRCh38
rs964857512 Chr.12: 110019027-110019027 on Build GRCh38
rs778682050 Chr.12: 110019069-110019069 on Build GRCh38
rs777439493 Chr.12: 110019174-110019174 on Build GRCh38
rs758371790 Chr.12: 110019011-110019011 on Build GRCh38
rs897154419 Chr.12: 110019059-110019059 on Build GRCh38
rs752473801 Chr.12: 110019108-110019108 on Build GRCh38
rs1012068764 Chr.12: 110019148-110019148 on Build GRCh38
rs748494840 Chr.12: 110019166-110019166 on Build GRCh38
rs759400191 Chr.12: 110019192-110019192 on Build GRCh38
rs781429761 Chr.12: 110019158-110019158 on Build GRCh38
rs774316993 Chr.12: 110019103-110019103 on Build GRCh38
rs770394460 Chr.12: 110019098-110019098 on Build GRCh38
rs975637811 Chr.12: 110019035-110019035 on Build GRCh38
rs758593632 Chr.12: 110019125-110019125 on Build GRCh38

View Full Product Details